Skip to Content
Merck
All Photos(2)

Key Documents

17-700

Sigma-Aldrich

Magna RIP® RNA-Binding Protein Immunoprecipitation Kit

Magna RIP® RNA-Binding Protein Immunoprecipitation Kit

RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.

Synonym(s):

Magnetic Bead RNA Immunoprecipitation, Magnetic RIP kit, RIP Kit, RNA Immunopreciptation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

manufacturer/tradename

Magna RIP®
Upstate®

technique(s)

RIP: suitable
immunoprecipitation (IP): suitable

shipped in

dry ice

Related Categories

General description

Gene regulation plays a critical role in complex cellular processes such as development, differentiation, and cellular response to environmental changes. In addition to transcriptional regulation of gene expression by transcription factors, cells utilize post-transcriptional regulatory mechanisms. One such mechanism involves use of certain RNA-binding proteins (RBPs) to temporally and coordinately regulate the rate of mRNA translation of functionally related gene
products. While the regulation of gene expression by transcription factors has been well studied over time, the post-transcriptional regulation of mRNAs by RBPs and the role of non-coding RNAs in this process is a relatively nascent field that remains to be thoroughly explored.
RNA-binding protein immunoprecipitation (RIP) is the RNA analog of the more well-known ChIP application (chromatin immunoprecipitation), which identifies DNA targets of DNA-binding proteins in an in-vivo cellular context. RIP can be used to identify specific RNA molecules (of many types) associated with specific nuclear or cytoplasmic binding proteins. These experiments involve immunoprecipitation of endogenously formed complexes of RNA-binding proteins and co-isolation of any RNA species associated with that RNA-binding protein. Purification of these RNA species allows interrogation and identification of mRNAs (and potentially non-coding RNAs associated with them) and can be directly measured using down stream applications including quantitative reverse transcription polymerase chain reaction (RT-PCR), microarray analysis (RIP-chip) and “deep-sequencing” or 2nd-generation sequencing based platforms (RIP-Seq).

Features & Benefits:
-Protein A/G magnetic beads, optimized to bind nucleic acid-protein immune complexes
-RNAse inhibitors and RNAse-free reagents
-Negative controls

Application

Research Category
Epigenetics & Nuclear Function

Packaging

RIP Kit capacity: 12 RNA-binding protein immunoprecipitation assays

Components

Magnetic Beads Protein A/G

RIP Wash Buffer

RIP Lysis Buffer

0.5 M EDTA

10% SDS

Salt Solution I

Salt Solution II

Precipitate Enhancer

Normal Mouse IgG

Rabbit IgG Purified

Protease Inhibitor Cocktail 200X

RNase Inhibitor

Proteinase K (10 mg/mL)

Nuclease free water

Physical form

Two boxes containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions.

Storage and Stability

Upon receipt, store components at the temperatures indicated on the labels. Kit components are stable for 6 months from date of shipment when stored as directed.

Legal Information

MAGNA RIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Downregulated LncRNA-ANCR promotes osteoblast differentiation by targeting EZH2 and regulating Runx2 expression.
Zhu, L; Xu, PC
Biochemical and biophysical research communications null
Active stabilization of human endothelial nitric oxide synthase mRNA by hnRNP E1 protects against antisense RNA and microRNAs.
Ho, JJ; Robb, GB; Tai, SC; Turgeon, PJ; Mawji, IA; Man, HS; Marsden, PA
Molecular and cellular biology null
Ribonomics: identifying mRNA subsets in mRNP complexes using antibodies to RNA-binding proteins and genomic arrays.
Tenenbaum, Scott A, et al.
Methods, 26, 191-198 (2002)
RNA processing and modification protein, carbon catabolite repression 4 (Ccr4), arrests the cell cycle through p21-dependent and p53-independent pathway.
Yi, X; Hong, M; Gui, B; Chen, Z; Li, L; Xie, G; Liang, J; Wang, X; Shang, Y
The Journal of Biological Chemistry null
Ubiquitination and deubiquitination of NP protein regulates influenza A virus RNA replication.
Liao, TL; Wu, CY; Su, WC; Jeng, KS; Lai, MM
The Embo Journal null

Questions

1–3 of 3 Questions  
  1. Is an antibody suitable for Co-IP also suitable for RIP? Is there any difference between hosts/immunoglobulin affinity?

    1 answer
    1. An antibody suitable for co-immunoprecipitation (Co-IP) of proteins is not necessarily suitable for RNA immunoprecipitation (RIP). Regardless of host, an antibody suitable for RIP targets accessible epitopes on the RNA binding protein of interest, which are not masked by the interaction with target RNAs. The amount of antibody used for RIP depends on the presentation (e.g., purified or unpurified) and the effective affinity of the candidate antibody when used for immunoprecipitation. It is recommended to use RIP-validated antibodies, such as RIPAb+ validated antibodies. Examples can be found at the following link: https://www.sigmaaldrich.com/search/ripab%2B-validated-antibody?focus=products&page=1&perpage=30&sort=relevance&term=RIPAb%2B%20validated%20antibody&type=product

      Helpful?

  2. For RIP-seq, should I perform rRNA depletion after RNA purification?

    1 answer
    1. Assuming rRNA are not known targets and the target RNA has been enriched using the binding protein/antibody, a depletion step is not required.

      Helpful?

  3. Hi, on the Magna RIP™ RNA-Binding Protein Immunoprecipitation Kit, after RNA insolation do RNA samples need DNAseI treatment to eliminate DNA contaminants? Or DNA was eliminated in a different way? Thanks Martin

    1 answer
    1. The need for DNA removal from RIPed RNA can vary depending on the specific RNA binding protein targets. Typically, most RIPed RNA samples do not require DNA removal. However, if DNA contamination of the sample is anticipated, it is recommended to perform DNase I treatment or include a minus RT (reverse transcription) sample for the qPCR analysis.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service