Skip to Content
Merck
All Photos(1)

Documents

20-400

Sigma-Aldrich

Magna GrIP Rack (8 well)

polyethylene rack, to hold, 15 mL (tubes), to hold, 0.5 mL (tubes), suitable for ChIP, suitable for RIP

Synonym(s):

Magnetic tube rack 0.5ml, Magnetic tube rack 15ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NA.41

material

polyethylene rack

Quality Level

feature

to hold 0.5 mL (tubes)
to hold 15 mL (tubes)

technique(s)

ChIP: suitable
RIP: suitable

shipped in

ambient

Related Categories

General description

A polyethylene rack containing 4 neodinium magnets used for ChIP and RIP assays. The rack is versatile & may also be used with either 15 ml or 0.5 ml tubes simply by standing the rack on either side.
The Magna GrIP Rack is a polyethylene rack containing 4 neodinium magnets. The rack is designed to rapidly and easily pellet samples bound to magnetic particles from the supernatant. In its horizontal position, the rack contains 8 holes suitable for 1.5-2.0 ml tubes or spin columns. The rack is versatile, and may also be used with either 15 ml or alternately, 0.5 ml tubes, simply by standing the rack on either side.

Application

Research Category
Cell Culture

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification and biochemical characterization of a putative [6Fe-6S] prismane-cluster-containing protein from Desulfovibrio vulgaris (Hildenborough).
A J Pierik,R B Wolbert,P H Mutsaers,W R Hagen,C Veeger
European Journal of Biochemistry null
Effect of a salmon diet on the distribution of plasma lipoproteins and apolipoproteins in normolipidemic adult men.
F T Lindgren,G L Adamson,V G Shore,G J Nelson,P C Schmidt
Lipids null
The amino acid sequence of rabbit J chain in secretory immunoglobulin A.
G J Hughes,S Frutiger,N Paquet,J C Jaton
The Biochemical Journal null
Open channel noise. V. Fluctuating barriers to ion entry in gramicidin A channels.
S H Heinemann,F J Sigworth
Biophysical Journal null
Determination of levamisole and thiabendazole in meat by HPLC and photodiode array detection.
B Wyhowski de Bukanski,J M Degroodt,H Beernaert
Zeitschrift fur Lebensmittel-Untersuchung Und -Forschung null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service