LSKMAGAG
PureProteome Protein A/G Mix Magnetic Beads
The PureProteome Protein A/G Mix Magnetic Bead System provides researchers with the power of the combination of both Protein A and Protein G immunoglobulin binding affinities in one convenient magnetic bead product.
Synonym(s):
PureProteome magnetic beads
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105507
eCl@ss:
32160405
NACRES:
NA.56
Recommended Products
packaging
pkg of 2 × 1 mL
manufacturer/tradename
PureProteome
technique(s)
immunoprecipitation (IP): suitable (crosslinking)
protein purification: suitable (antibody)
particle size
10 μm
capacity
2-3 mg/mL, suspension binding capacity (rabbit IgG)
shipped in
wet ice
General description
The PureProteome Protein A/G Mix Magnetic Bead System provides researchers with the power of the combination of both Protein A and Protein G immunoglobulin binding affinities in one convenient magnetic bead product, resulting in greater antibody compatibility than either Protein A or Protein G alone.
Application
PureProteome™ Protein A/G Mix Magnetic Beads have been used in:
- immunoprecipitation
- crosslinking immunoprecipitation
- antibody purification; protein purification
- co‑immunoprecipitation (Co‑IP) assay
Features and Benefits
- Offers a rapid and reproducible method for immunoglobulin purification from complex mixtures such as serum, plasma, or cell culture supernatant samples.
- A magnetic rack or platform is used to capture the beads from wash and elution fractions during antibody purification.
- Preferred for low throughput using single microfuge tubes or higher throughput using automation.
Legal Information
PureProteome is a trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service