Skip to Content
Merck
All Photos(1)

Key Documents

1.46813

Millipore

Luria Broth

bottle capacity × filling volume 250 mL × 200 mL, closure type, screw cap with 3 loci

Synonym(s):

LB Broth, E. coli enrichment broth, LB broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225

Agency

EP 2.6.1
JP 4.06
USP 71
71

sterility

sterile; autoclaved

form

liquid

feature

closure type screw cap with 3 loci
ready-to-use

packaging

bottle of

technique(s)

direct inoculation: suitable

bottle capacity × filling volume

250 mL × 200 mL

pH

7.0 ± 0.2

application(s)

cosmetics
food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

selective for bacteria: Escherichia Coli

Looking for similar products? Visit Product Comparison Guide

General description

Luria Broth is a nutrient rich media designed for the cultivation of E. coli in samples from food and other materials.
The medium is supplemented with casein and yeast extract supplying essential nutrients for growth. The presence of essential electrolytes in the medium maintains osmotic balance and promotes growth. Selective cultivation is achieved on adding specific antibiotics.

Application

Luria Broth is used to cultivate and promote growth of E. coli in test samples from the food and beverage industry.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service