Skip to Content
Merck

1.37136

SAFC

Tetrahydrofuran

EMPROVE® ESSENTIAL

Pharma Manufacturing

Synonym(s):

Tetrahydrofuran, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
MDL number:
UNSPSC Code:
12352005

vapor density

2.5 (vs air)

Quality Level

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

product line

EMPROVE® ESSENTIAL

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

610 °F

expl. lim.

1.8-11.8 %

technique(s)

API processing | polymorph screening: suitable

refractive index

n20/D 1.407 (lit.)

pH

7-8 (20 °C, 200 g/L in H2O)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

storage temp.

2-25°C

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Tetrahydrofuran is a versatile organic solvent for API synthesis, as a pre-cursor to polymers and it is used for several extended applications during drug manufacturing. The performance of the processes increases with the usage of Tetrahydrofuran in certain API′s than other available solvents. Tetrahydrofuran Emprove® Essential is manufactured according to GMP guidelines and is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service