Skip to Content
Merck
All Photos(2)

Key Documents

34865

Sigma-Aldrich

Tetrahydrofuran

≥99.9%, suitable for HPLC, inhibitor-free

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Product Name

Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%

grade

HPLC grade

Quality Level

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

610 °F

expl. lim.

1.8-11.8 %

technique(s)

HPLC: suitable

impurities

≤0.0005% non-volatile matter
≤0.002% free acid (as CH3COOH)
≤0.05% peroxides (as H2O2)
≤0.05% water (Karl Fischer)

evapn. residue

≤0.0005%

color

APHA: ≤10

refractive index

n20/D 1.407 (lit.)

pH

~7

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

absorption

≤0.0044 at 315 nm
≤0.005 at 350 nm
≤0.005 at 400 nm
≤0.02 at 300 nm
≤0.046 at 275 nm
≤0.18 at 250 nm
≤0.26 at 245 nm
≤1.0 at 212 nm

UV absorption

λ: 245 nm Amax: ≤0.26
λ: 275 nm Amax: ≤0.046
λ: 315 μm Amax: ≤0.0044

suitability

complies for IR spectroscopy

application(s)

food and beverages

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofuran (THF) is widely employed as a solvent. It constitutes the key fragment of various natural products (polyether antibiotics). THF forms a double hydrate with hydrogen sulfide. Crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies. Butane-1,4-diol is formed as an intermediate during the synthesis of THF. Hot THF is useful for the dissolution of polyvinylidene chloride (PVDV).

Application

Suitable for HPLC, spectrophotometry, environmental testing
Tetrahydrofuran may be used for the dissolution of poly-ε-caprolactone (PCL) and 1,3-diaminopentane, during the preparation of poly-ε-caprolactone (PCL)-hydroxyapatite (HA) scaffolds and acrylate-terminated poly(5-amino-1-pentanol-co-1,4-butanediol diacrylate) (C32)- 1,3-diaminopentane (117) polymer, respectively.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Other Notes

Important notice
  • The article number 34865-4X2.5L will be discontinued. Please order the single bottle 34865-2.5L which is physically identical with the same exact specifications.
  • The article number 34865-6X1L will be discontinued. Please order the single bottle 34865-1L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd J Harris et al.
Biomaterials, 31(5), 998-1006 (2009-10-24)
The use of biomaterials for gene delivery can potentially avoid many of the safety concerns with viral gene delivery. However, the efficacy of polymeric gene delivery methods is low, particularly in vivo. One significant concern is that the interior and
Eagleson M.
Concise Encyclopedia Chemistry, 883-883 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 152-152 (1994)
Synthetic routes to tetrahydrofuran, tetrahydropyran, and spiroketal units of polyether antibiotics and a survey of spiroketals of other natural products.
Boivin TLB.
Tetrahedron, 43(15), 3309-3362 (1987)
Soni Khandelwal et al.
International journal of molecular sciences, 19(11) (2018-10-31)
Within the subtypes of breast cancer, those identified as triple negative for expression of estrogen receptor α (ESR1), progesterone receptor (PR) and human epidermal growth factor 2 (HER2), account for 10⁻20% of breast cancers, yet result in 30% of global

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service