Skip to Content
Merck
All Photos(1)

Key Documents

124018

Sigma-Aldrich

Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2

Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2, CAS 612847-09-3, is a cell-permeable, reversible inhibitor of Akt1/Akt2 (IC₅₀ = 58 nM, 210 nM, & 2.12 µM for Akt1, Akt2, and Akt3, respectively).

Synonym(s):

Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2, 1,3-Dihydro-1-(1-((4-(6-phenyl-1H-imidazo[4,5-g]quinoxalin-7-yl)phenyl)methyl)-4-piperidinyl)-2H-benzimidazol-2-one, Akti-1/2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H29N7O
CAS Number:
Molecular Weight:
551.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N6(CCC(CC6)[n]7c8c(nc7O)cccc8)Cc1ccc(cc1)c2nc3c(nc2c5ccccc5)cc4[nH]cnc4c3

InChI

1S/C34H29N7O/c42-34-39-26-8-4-5-9-31(26)41(34)25-14-16-40(17-15-25)20-22-10-12-24(13-11-22)33-32(23-6-2-1-3-7-23)37-29-18-27-28(36-21-35-27)19-30(29)38-33/h1-13,18-19,21,25H,14-17,20H2,(H,35,36)(H,39,42)

InChI key

BIWGYFZAEWGBAL-UHFFFAOYSA-N

General description

A cell-permeable and reversible quinoxaline compound that potently and selectively inhibits Akt1/Akt2 activity (IC50 = 58 nM, 210 nM, and 2.12 µM for Akt1, Akt2, and Akt3, respectively, in in vitro kinase assays). The inhibition appears to be pleckstrin homology (PH) domain-dependent. It does not exhibit any inhibitory effect against PH domain-lacking Akts, or other closely related AGC family kinases, PKA, PKC, and SGK, even at concentrations as high as 50 µM. Overcomes Akt1/Akt2-mediated resistance to chemotherapeutics in tumor cells and is shown to block basal and stimulated phosphorylation/activation of Akt1/Akt2 both in cultured cells in vitro and in mice in vivo. A 10 mM (1 mg/181 µl) solution of Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 (Cat. No. 124017) in DMSO is also available.
A cell-permeable, reversible, potent, and selective inhibitor of Akt1/Akt2 activity (IC50 = 58 nM, 210 nM, and 2.12 µM for Akt1, Akt2, and Akt3, respectively, in in vitro kinase assays). The inhibition is reported to be pleckstrin homology (PH) domain-dependent. It does not exhibit any inhibitory effect against PH domain-lacking Akts, or other closely related AGC family kinases, PKA, PKC, and SGK, even at concentrations as high as 50 µM. Overcomes Akt1/Akt2-mediated resistance to chemotherapeutics in tumor cells and is shown to block basal and stimulated phosphorylation/activation of Akt1/Akt2 both in cultured cells in vitro and in mice in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Akt1, Akt2, Akt3
Product does not compete with ATP.
Reversible: yes
Target IC50: 58 nM, 210 nM, 2.12 µM against Akt1, Akt2, and Akt3, respectively, in in vitro kinase assays

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C.

Other Notes

Calleja, V., et al. 2009. PLoS Biol.7, e17.
Logie, L., et al. 2007. Diabetes56, 228.
Barnett, S.F., et al. 2005. Biochem. J.385, 399.
DeFeo-Jones, D., et al. 2005. Mol. Cancer Ther.4, 271.
Zhao, Z., et al. 2005. Bioorg. Med. Chem. Lett.15, 905.
Lindsley, C.W., et al. 2005. Bioorg. Med. Chem. Lett.15, 761.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriel Luta et al.
International journal of molecular sciences, 22(8) (2021-05-01)
In the latest years, there has been an increased interest in nanomaterials that may provide promising novel approaches to disease diagnostics and therapeutics. Our previous results demonstrated that Carbon-dots prepared from N-hydroxyphthalimide (CD-NHF) exhibited anti-tumoral activity on several cancer cell
Giacomo Della Verde et al.
Life science alliance, 5(3) (2021-12-15)
Fms-like tyrosine kinase 3 (Flt3) is a regulator of hematopoietic progenitor cells and a target of tyrosine kinase inhibitors. Flt3-targeting tyrosine kinase inhibitors can have cardiovascular side effects. Flt3 and its ligand (Flt3L) are expressed in the heart, but little
M L Brown Lobbins et al.
Clinical and experimental immunology, 191(1), 84-95 (2017-09-09)
Peripheral blood mononuclear cells taken from patients with scleroderma express increased levels of interleukin (IL)-13. Moreover, the expression of matrix metalloproteinase-1 (MMP-1) from involved scleroderma skin fibroblasts is refractory to stimulation by tumour necrosis factor (TNF)-α. To elucidate the mechanism(s)
Jianbin Su et al.
Frontiers in pharmacology, 13, 857081-857081 (2022-04-05)
Microglia are key components of the central innate immune system. The over-activation of microglia, which occurs in nervous system disorders, is usually accompanied with retractions of their ramified processes. Reversing of microglial process retraction is a potential strategy for the
Lilija Wehling et al.
eLife, 11 (2022-10-19)
The Hippo signaling pathway controls cell proliferation and tissue regeneration via its transcriptional effectors yes-associated protein (YAP) and transcriptional coactivator with PDZ-binding motif (TAZ). The canonical pathway topology is characterized by sequential phosphorylation of kinases in the cytoplasm that defines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service