Skip to Content
Merck
All Photos(2)

Key Documents

516535

Sigma-Aldrich

GSK2606414

≥99% (HPLC), powder, PERK inhibitor, Calbiochem®

Synonym(s):

PERK Inhibitor I, GSK2606414

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20F3N5O
CAS Number:
Molecular Weight:
451.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

PERK Inhibitor I, GSK2606414, GSK2606414 is a cell-permeable, highly potent inhibitor of EIF2AK3/PERK (IC₅₀ = 0.4 nM; [ATP] = 5 µM). Targets PERK in its inactive DFG conformation at the ATP-binding region.

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CC1=CC=CC(C(F)(F)F)=C1)N2CCC3=C2C=CC(C4=CN(C)C5=C4C(N)=NC=N5)=C3

InChI

1S/C24H19F4N5O/c1-32-11-18(21-22(29)30-12-31-23(21)32)14-2-3-19-15(9-14)4-5-33(19)20(34)8-13-6-16(24(26,27)28)10-17(25)7-13/h2-3,6-7,9-12H,4-5,8H2,1H3,(H2,29,30,31)

InChI key

PXVQGBJMIQCDEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A cell-permeable pyrrolopyrimidamine compound that acts as a highly potent EIF2AK3/PERK inhibitor (IC50 = 0.4 nM; [ATP] = 5 µM) by targeting PERK in its inactive DFG conformation at the ATP-binding region, while displaying ≥385-fold selectivity over c-Kit, Aurora B, BRK, HRI/EIF2AK1, MLK2/MAP3K10, c-MER, DDR2, PKR/EIF2AK2, and MLCK2/MYLK2 (IC50 = 154, 407, 412, 420, 452, 474, 524, 696, and 701 nM, respectively) and little activity against more than 280 other kinases (IC50 >1 µM). Shown to block ER stress-induced PERK autophosphorylation following Thapsigargin addition in A549 cultures in vitro (by 100% at ≤30 nM; 60 min preincubation) and effectively retard PxBC-3 tumor growth in mice in vivo (50 and 150 mg/kg/12 h p.o.). Also available as a 25 mM solution in DMSO (Cat. No. 508340).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Perk
Reversible: yes
Target IC50: 0.4 nM against EIF2AK3/PERK-catalyzed EIF2&alpha

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liliana Matos et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 70(8), 924-935 (2014-08-26)
The aging process is characterized by progressive accumulation of damaged biomolecules in the endoplasmic reticulum, as result of increased oxidative stress accompanying cellular senescence. In agreement, we hypothesized that WI-38 human cellular models of replicative senescence and stress-induced premature senescence
Bibha Dahal et al.
Virology, 539, 121-128 (2019-11-17)
Venezuelan equine encephalitis virus (VEEV) is a neurotropic virus that causes significant disease in both humans and equines. Here we characterized the impact of VEEV on signaling pathways regulating cell death in human primary astrocytes. VEEV productively infected primary astrocytes
Daniel R Sandoval et al.
bioRxiv : the preprint server for biology (2021-04-02)
We identify the prolyl-tRNA synthetase (PRS) inhibitor halofuginone 1 , a compound in clinical trials for anti-fibrotic and anti-inflammatory applications 2 , as a potent inhibitor of SARS-CoV-2 infection and replication. The interaction of SARS-CoV-2 spike protein with cell surface
Kai Chen et al.
Aging cell, 21(4), e13592-e13592 (2022-03-18)
Delirium is the most common postoperative complication in older patients after prolonged anesthesia and surgery and is associated with accelerated cognitive decline and dementia. The neuronal pathogenesis of postoperative delirium is largely unknown. The unfolded protein response (UPR) is an
Peter van Galen et al.
Nature, 510(7504), 268-272 (2014-04-30)
The blood system is sustained by a pool of haematopoietic stem cells (HSCs) that are long-lived due to their capacity for self-renewal. A consequence of longevity is exposure to stress stimuli including reactive oxygen species (ROS), nutrient fluctuation and DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service