Skip to Content
Merck
All Photos(1)

Key Documents

1.14434

Supelco

Chlorine Test Kit, colorimetric

0.010-0.30 mg/L (Cl₂), MQuant®

Synonym(s):

Chlorine Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Chlorine Test, colorimetric, 0.010-0.30 mg/L (Cl2), for use with MQUANT®

product line

MQuant®

Quality Level

specific analyte(s)

chlorine

measuring range

0.010-0.30 mg/L (Cl2)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

(free chlorine) Method: colorimetric, DPD, with color card and comparator block with long test tubes 0.010 - 0.025 - 0.045 - 0.06 - 0.08 - 0.10 - 0.15 - 0.20 - 0.30 mg/l Cl₂ MQuant®
The MQuant® Chlorine test (formally MColortest®) can be used for the determination of free chlorine.

The test can be used in
  • Drinking water and mineral water
  • Waters from aquaculture
  • Wastewater
  • Electroplating wastewater
  • Disinfectant solutions

This test is not suited for seawater.

Method
In weakly acidic solution free chlorine reacts with diethyl-p-phenylenediamine (DPD) to form a red-violet dye. The chlorine concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card.
The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range. Due to the long test tubes a sensitive measurement range can be covered. Using the refill pack 1.14977 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system. 

Application

  • 3,3′,5,5′-tetramethylbenzidine as multi-colorimetric indicator of chlorine in water in line with health guideline values.: This study demonstrates the use of 3,3′,5,5′-tetramethylbenzidine as a colorimetric indicator for detecting chlorine levels in water, providing a cost-effective and simple method for ensuring water safety in compliance with health guidelines (Palladino et al., 2020).
  • Analysis of free chlorine in aqueous solution at very low concentration with lateral flow tests.: Investigates the effectiveness of lateral flow assays for the quantification of free chlorine in water, highlighting their potential for quick and accurate chlorine testing in field settings (Schwenke et al., 2019).
  • Improved residual chlorine test; the ortho-toluidine: arsenite method for testing water for true, false and chloramine residuals.: Describes a methodological advancement in measuring different forms of chlorine residuals in water, enhancing the accuracy of water quality assessments (GILCREAS FW, 1948).

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service