1.00599
Chlorine Test, photometric
photometric, 0.010-6.00 mg/L (Cl2), Spectroquant®
About This Item
Recommended Products
product name
Chlorine Test, photometric, 0.010-6.00 mg/L (Cl2), Spectroquant®
product line
Spectroquant®
Quality Level
usage
sufficient for 200 tests ((free chlorine) or 100 (free chlorine) + 100 (total chlorine))
specific analyte(s)
chlorine
measuring range
0.010-6.00 mg/L (Cl2)
technique(s)
photometry: suitable
compatibility
for use with Spectroquant® Move 100
for use with Spectroquant® Move DC
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600
detection method
photometric (DPD)
storage temp.
15-25°C
General description
This Spectroquant® Chlorine test allows the accurate quantification. Chlorinated disinfectants are still in common use all over the world, their major applications being in the chlorination of drinking water and swimming pools. In weakly acidic solution free chlorine reacts with dipropyl-p-phenylenediamine (DPD) to form a red-violet dye that is determined photometrically. In the presence of potassium iodide, also combined chlorine is measured in this reaction.
The method is analogous to EPA 330.5, APHA 4500-Cl2 G, and DIN EN ISO 7393-2.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.
Application
- 3,3′,5,5′-tetramethylbenzidine as multi-colorimetric indicator of chlorine in water in line with health guideline values.: This study demonstrates the use of 3,3′,5,5′-tetramethylbenzidine as a colorimetric indicator for detecting chlorine levels in water, providing a cost-effective and simple method for ensuring water safety in compliance with health guidelines (Palladino et al., 2020).
- Analysis of free chlorine in aqueous solution at very low concentration with lateral flow tests.: Investigates the effectiveness of lateral flow assays for the quantification of free chlorine in water, highlighting their potential for quick and accurate chlorine testing in field settings (Schwenke et al., 2019).
- Improved residual chlorine test; the ortho-toluidine: arsenite method for testing water for true, false and chloramine residuals.: Describes a methodological advancement in measuring different forms of chlorine residuals in water, enhancing the accuracy of water quality assessments (GILCREAS FW, 1948).
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Repr. 1B - STOT RE 2 Oral
Target Organs
Thyroid
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service