Skip to Content
Merck
All Photos(1)

Key Documents

1.09743

Supelco

Methyl oleate

reference substance for gas chromatography

Synonym(s):

Methyl oleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O2
CAS Number:
Molecular Weight:
296.49
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-992-5
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

form

liquid

potency

>2000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

bp

202-210 °C/20 hPa

mp

-20 °C

transition temp

flash point 113 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10+

InChI key

QYDYPVFESGNLHU-ZHACJKMWSA-N

Analysis Note

Purity (GC): ≥ 96.0
Identity (IR-spectrum): conforms
methyl elaidate (GC): ≤ 1.0
Methyl linoleate (GC): ≤ 1.0
Methyl stearate (GC): ≤ 1.0

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service