Skip to Content
Merck
All Photos(4)

Key Documents

P5177

Sigma-Aldrich

Methyl palmitate

≥99% (capillary GC)

Synonym(s):

n-Hexadecanoic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CO2CH3
CAS Number:
Molecular Weight:
270.45
Beilstein:
1780973
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

plant (palm)

Assay

≥99% (capillary GC)

form

solid

refractive index

n20/D 1.4512 (lit.)

bp

185 °C/10 mmHg (lit.)

mp

32-35 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C17H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h3-16H2,1-2H3

InChI key

FLIACVVOZYBSBS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Methyl palmitate (PAME), a fatty acid ester of palmitic acid, is a muscurinic receptors antagonist.[1] Methyl palmitate functions as a vasodilator and relaxes the arterioles in retina upon electrical depolarization.[2] In the superior cervical ganglion, methyl palmitate modulates nicotinic receptor.[3] Endogenous methyl palmitate modulates nicotinic receptor-mediated transmission in the superior cervical ganglion.[3] PAME activates Kv7 channels and promotes perivascular adipose tissue.[4]

Application

Methyl palmitate has been used to test impact of cerebral blood flow using laser doppler flowmetry (LDF) experiments [5] and as a standard to determine retention indices (RIs) using gas chromatograph−mass spectrometer[6]

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palmitic acid methyl ester and its relation to control of tone of human visceral arteries and rat aortas by perivascular adipose tissue
Wang N, et al.
Frontiers in Physiology, 9 (2018)
Terminal double bonds in the alkenes and acetates of defensive secretion from the thrips Suocerathrips linguis Mound & Marullo, 1994 (Thysanoptera: Phlaeothripidae)
Gehlsen U, et al.
Chemoecology, 19(2), 97-102 (2009)
Jiao Liu et al.
Biotechnology for biofuels, 8, 229-229 (2016-01-01)
Fatty acid composition is an important physiological parameter of microalgae, which is taken as the third generation alternative resource of biodiesel. To boost microalgal research and applications, a convenient, rapid, and acid-catalyzed transesterification procedure that satisfies the demand for the
Endogenous methyl palmitate modulates nicotinic receptor-mediated transmission in the superior cervical ganglion
Lin HW, et al.
Proceedings of the National Academy of Sciences of the USA, 1051(49), 19526-19531 (2008)
Palmitic acid methyl ester is a novel neuroprotective agent against cardiac arrest
Lee RHC, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids (2018)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service