Skip to Content
Merck
All Photos(1)

Documents

1.04335

Supelco

Isohexane

≥99.0% (GC), suitable for HPLC

Synonym(s):

Isohexane, 2-Methyl pentane, iso-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
UNSPSC Code:
12191502
EC Index Number:
295-570-2
NACRES:
NA.03

product name

Isohexane, for liquid chromatography LiChrosolv®

vapor pressure

160-190 hPa ( 20 °C)

Quality Level

product line

LiChrosolv®

grade

isocratic

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

260 °C

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.0-7.4 % (v/v)

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.005% Water
≤0.5% n-Hexane (GC)
≥60% Isohexane (GC)

evapn. residue

≤2.0 mg/L

color

APHA: ≤10

transmittance

210 nm, ≥60%
220 nm, ≥80%
245 nm, ≥98%

kinematic viscosity

0.5 cSt(20 °C)

bp

53-63 °C/1013 hPa

mp

<-50 °C

transition temp

flash point -21 °C

density

0.66 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Application


  • In-house validation of accelerated solvent extraction-gas chromatography-mass spectrometry for the determination of bound 3- and 2-monochloropropanediols (MCPD) and glycidol in food products.: This study showcases the validation of a robust analytical method, crucial for ensuring food safety and compliance with regulations. The use of isohexane in the process highlights its application in improving extraction efficiency and analysis precision (Shaari et al., 2021).

  • Optimized determination of polybrominated diphenyl ethers and polychlorinated biphenyls in sheep serum by solid-phase extraction-gas chromatography-mass spectrometry.: Demonstrates the refinement of extraction and detection techniques using isohexane for environmental pollutants in biological samples, enhancing the sensitivity and accuracy of these measurements (Zhang & Rhind, 2011).

  • Simultaneous extraction and clean-up of polybrominated diphenyl ethers and polychlorinated biphenyls from sheep liver tissue by selective pressurized liquid extraction and analysis by gas chromatography-mass spectrometry.: This article describes a streamlined method to simultaneously extract and purify specific contaminants from liver tissue, with isohexane playing a critical role in the efficiency of the extraction process (Zhang et al., 2011).

  • Selective pressurized liquid extraction of estrogenic compounds in soil and analysis by gas chromatography-mass spectrometry.: Focuses on the targeted extraction of hormone disruptors from environmental samples, where isohexane′s selective solvation properties are utilized to enhance the selectivity and yield of the analytes (Zhang et al., 2011).

  • Fast sample preparation involving MASE and coupled column normal phase liquid chromatography for the rapid trace analysis of dioxins in air-dust samples from fire catastrophe emissions.: Discusses a rapid preparation and analysis method for trace contaminants in complex matrices, underscoring isohexane′s role in achieving fast and reliable results (van Beuzekom et al., 2004).



Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity C₆H₁₄-isomeres (GC): ≥ 99.0 %
Isohexane (GC): ≥ 60.0 %
n-Hexane (GC): ≤ 0.5 %
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.005 %
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 210 nm): ≥ 60 %
Transmission (at 220 nm): ≥ 80 %
Transmission (from 245 nm): ≥ 98 %
Filtered by 0.2 µm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-5.8 °F

Flash Point(C)

-21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service