Skip to Content
Merck
All Photos(2)

Documents

1.00868

Supelco

Ethyl acetate

for liquid chromatography LiChrosolv®

Synonym(s):

Ethyl ethanoate, Acetic ester, Ethyl ester, EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
12191502
EC Index Number:
205-500-4
NACRES:
NA.03

grade

HPLC grade
isocratic

Quality Level

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

product line

LiChrosolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

801 °F

potency

5620 mg/kg LD50, oral (Rat)
>18000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.05% Water

evapn. residue

≤2.0 mg/L

transmittance

260 nm, ≥50%
265 nm, ≥80%
270 nm, ≥98%

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

transition temp

flash point -4 °C

density

0.902 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Application


  • [Determination of 12 prohibited veterinary drug residues in pig urine by ultra high performance liquid chromatography-tandem mass spectrometry]: Ethyl acetate is utilized for sample preparation in this advanced method to detect drug residues in animal samples, emphasizing its role in veterinary pharmaceutical compliance (Wan et al., 2024).

  • Therapeutic effect of Ginkgetin on smoke-induced airway inflammation by down-regulating the c/EBPβ signaling pathway and CCL2 expression: This study explores the anti-inflammatory properties of Ginkgetin, where ethyl acetate is used in the extraction process, highlighting its utility in biomedical research (Tao et al., 2024).

  • Effect of methanol extract of Plectranthus esculentus N.E.Br tuber and its fractions on indices of benign prostatic hyperplasia in Wistar rats: Ethyl acetate is employed in the extraction of bioactive compounds for pharmacological studies, showing its critical role in therapeutic applications (Kanu et al., 2024).

  • Phytochemical evaluation of Ziziphus mucronata and Xysmalobium undulutum towards the discovery and development of anti-malarial drugs: Demonstrates the use of ethyl acetate in phytochemical screenings, contributing to the development of new anti-malarial medications (Buthelezi et al., 2024).

  • The First Records of the In Silico Antiviral and Antibacterial Actions of Molecules Detected in Extracts of Algerian Fir (Abies numidica De Lannoy) Using LC-MS/MS Analysis: This research utilizes ethyl acetate for extracting compounds from plants for antiviral and antibacterial testing, underscoring its importance in the study of natural products (Benouchenne et al., 2024).

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service