Skip to Content
Merck
All Photos(1)

Key Documents

1.00675

Supelco

AOX Cell Test

photometric, 0.05-2.50 mg/L (AOX), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

analyte functional class(es)

adsorbable organic halides (AOX)

measuring range

0.05-2.50 mg/L (AOX)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Iron(III)-thiocyanate)

storage temp.

15-25°C

General description

Method: photometric 0.05 - 2.50 mg/l AOX Spectroquant®

Organic halogen compounds are adsorbed to activated charcoal and subsequently digested after any adhering inorganic halides are washed out. The resultant halide is then determined photometrically.

The adsorption method is analogous to EN ISO 9562.

The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
For sample preparation and AOX enrichment it is required to purchase the Spectroquant® AOX Sample Preparation Set 1.00677 as well as the Spectroquant® Enrichment Set 1.00678 together with the Cell Test.


All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode), which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT RE 2

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - (Dimethylsulfoxide)

Flash Point(C)

87 °C - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service