Skip to Content
Merck
All Photos(2)

Key Documents

1.14878

Supelco

TOC Test, photometric

photometric, 5.0-80.0 mg/L (TOC), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

TOC Cell Test, photometric, 5.0-80.0 mg/L (TOC), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

analyte chemical class(es)

total organic carbon (TOC)

measuring range

5.0-80.0 mg/L (TOC)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Indicator)

storage temp.

15-25°C

General description

Method: photometric 5.0 - 80.0 mg/l Spectroquant®
This Spectroquant TOC test allows the accurate quantification of the TOC especially in sewage outflow samples. The term TOC (Total Organic Carbon) is used to describe the total content of organically bound carbon in dissolved and undissolved forms. By digestion with sulfuric acid and peroxodisulfate, these compounds are transformed into carbon dioxide.
This reacts with an indicator, the color is determined photometrically. Inorganic carbon (dissolved carbon dioxide and carbonates) is expelled beforehand by acidification.
The digestion is analogous to APHA 5310 C (2014).


All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.


The Spectroquant Cell Tests come with prefilled 16 mm round cells as well as all the required reagents to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Met. Corr. 1 - Ox. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis A Cueto et al.
Environmental science and pollution research international, 27(14), 15984-15993 (2019-05-06)
Phosphorus (P) is a nutrient for plant growth but also a pollutant in water bodies causing eutrophication. The source of P is mainly human and animal wastewater and runoffs from different land uses. The objective of the present study is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service