Skip to Content
Merck
All Photos(1)

Key Documents

1.00856

Supelco

Phenol Test, photometric

photometric, 0.002-0.100 mg/L (phenol), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.21

Product Name

Phenol Test, photometric, 0.002-0.100 mg/L (phenol), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 50-250 tests

specific analyte(s)

phenol

measuring range

0.002-0.100 mg/L (phenol)
0.025-5.0 mg/L (phenol)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (4-Aminoantipyrine)

storage temp.

15-25°C

General description

Method: photometric 0.002 - 5.00 mg/l Spectroquant®

The method is analogous to EPA 420.1, APHA 5530 C+D, ISO 6439, and ASTM D1783-01.

This Spectroquant®Phenol Reagent Test allows the accurate quantification of the phenol content (and its derivatives) in aqueous samples.

Method applied: In buffered solution, in the presence of an oxidizing agent phenol and its ortho- and meta-substituted compounds react with 4-aminoantipyrine to form a red compound which is determined photometrically.

The Spectroquant®Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Application

  • KUNKEL PHENOL TEST IN DIAGNOSIS OF ATHEROSCLEROSIS.: This article explores the utilization of the Kunkel phenol test in diagnosing atherosclerosis, indicating its relevance in clinical diagnostics rather than analytical chemistry applications (MEDVEDEV VP et al., 1964).

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service