Skip to Content
Merck
All Photos(1)

Key Documents

05-504

Sigma-Aldrich

Anti-IQGAP1 Antibody, clone AF4

clone AF4, Upstate®, from mouse

Synonym(s):

Anti-HUMORFA01, Anti-SAR1, Anti-p195

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

AF4, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IQGAP1(8826)

Specificity

IQGAP1, not IQGAP2

Immunogen

A Glu-Glu tagged C-terminal fragment corresponding to amino acids 863-1657 of human IQGAP1, purified from baculovirus infected insect cells

Application

Detect IQGAP1 using this Anti-IQGAP1 Antibody, clone AF4 validated for use in IP & WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

routinely evaluated by immunoblot on HepG2 RIPA cell lysate

Target description

185kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mireille Lambert et al.
The Journal of cell biology, 157(3), 469-479 (2002-04-24)
Cadherin receptors are key morphoregulatory molecules during development. To dissect their mode of action, we developed an approach based on the use of myogenic C2 cells and beads coated with an Ncad-Fc ligand, allowing us to mimic cadherin-mediated adhesion. We
Yuehong Cui et al.
OncoTargets and therapy, 10, 627-643 (2017-02-23)
In recent years, understanding of the role of asparaginyl endopeptidase (AEP) in tumorigenesis has steadily increased. In this study, we investigated whether AEP expression correlates with sensitivity to chemotherapeutic drugs in gastric cancer and explored the mechanism. AEP expression in
Martina Sanlorenzo et al.
Scientific reports, 8(1), 10902-10902 (2018-07-22)
Despite major advances in targeted melanoma therapies, drug resistance limits their efficacy. Long noncoding RNAs (lncRNAs) are transcriptome elements that do not encode proteins but are important regulatory molecules. LncRNAs have been implicated in cancer development and response to different
IQGAP1 involvement in MTOC and granule polarization in NK-cell cytotoxicity.
Namita Kanwar,John A Wilkins
European Journal of Immunology null
Katherine L Jameson et al.
Nature medicine, 19(5), 626-630 (2013-04-23)
Upregulation of the ERK1 and ERK2 (ERK1/2) MAP kinase (MAPK) cascade occurs in >30% of cancers, often through mutational activation of receptor tyrosine kinases or other upstream genes, including KRAS and BRAF. Efforts to target endogenous MAPKs are challenged by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service