Skip to Content
Merck
All Photos(1)

Key Documents

S2442

Sigma-Aldrich

S9 from Liver, Pooled

from human

Synonym(s):

Liver S9 Fraction, S9 Liver Extract

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501
NACRES:
NA.47

biological source

human

Quality Level

packaging

vial of ~20 mg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RPS9(6203)

Looking for similar products? Visit Product Comparison Guide

Application

Pooled S9 fractions from liver have been used for genotoxicity experiments.

Biochem/physiol Actions

Post-mitochondrial supernatant fraction from homogenized liver. Source of drug metabolizing enzymes including P-450, these pools are useful in the study of xenobiotic metabolism and drug interactions.
The hepatic S9 pools from a variety of biological sources represent the post-mitochondrial supernatant fraction from homogenized liver. Known to be a rich source of drug metabolizing enzymes including P-450, these pools are useful in the study of xenobiotic metabolism and drug interactions.
The hepatic S9 pools from a variety of biological sources represent the post-mitochondrial supernatant fraction from homogenized liver. They are known to be a rich source of drug metabolizing enzymes including P-450. S9 pools are useful in the study of xenobiotic metabolism and drug interactions. They are used in the Ames test, which is a method that uses bacteria to test if a given chemical causes mutations in the genetic material of a given test organism.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Ames Salmonella/microsome mutagenicity assay.
Mortelmans K and Zeiger E
Mutation Research (2000)
A conjugate of pyridine-4-aldoxime and atropine as a potential
antidote against organophosphorus compounds poisoning
Jasna Lovric
Acta Biochimica Polonica, 58(2) (2011)
Fabien Schultz et al.
Frontiers in pharmacology, 12, 678535-678535 (2021-07-20)
We investigated the potential antimalarial and toxicological effects of 16 medicinal plants frequently used by traditional healers to treat malaria, fever, and related disorders in the Greater Mpigi region in Uganda. Species studied were Albizia coriaria, Cassine buchananii, Combretum molle
Marina Paul et al.
Reproductive sciences (Thousand Oaks, Calif.) (2023-02-11)
The mechanism by which human labor is initiated in the presence of elevated circulating progesterone levels remains unknown. Recent evidence indicates that the progesterone-metabolizing enzyme, 20α-hydroxysteroid dehydrogenase (20α-HSD), encoded by the gene AKR1C1, may contribute to functional progesterone withdrawal. We
Implementation of the Three Rs in the Human Hazard
Assessment of Brazilian Medicinal Plants: An Evaluation
of the Cytotoxic and Genotoxic Potentials of Dipteryx
alata Vogel
Natalia M. Esteves-Pedro
Alberta RN / Alberta Association of Registered Nurses (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service