Skip to Content
Merck
All Photos(5)

Key Documents

05-101

Sigma-Aldrich

Anti-EGFR Antibody, neutralizing, clone LA1

clone LA1, Upstate®, from mouse

Synonym(s):

Anti-ERBB, Anti-ERBB1, Anti-ERRP, Anti-HER1, Anti-NISBD2, Anti-PIG61, Anti-mENA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LA1, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
neutralization: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)

General description

Epidermal growth factor receptor (UniProt: P00533; also known as EC:2.7.10.1, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1) is encoded by the EGFR (also known as ERBB, ERBB1, HER1) gene (Gene ID: 1956) in human. EGFR is a single-pass type I membrane glycoprotein that is ubiquitously expressed. It is synthesized with a signal peptide (1-24), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 25-645), a transmembrane domain (aa 646-668), and a long cytoplasmic domain (669-1210). It possesses receptor tyrosine kinase activity and upon binding of ligands activates several signaling cascades. Ligand binding is known to trigger receptor homo- and/or heterodimerization, which stimulates its intrinsic intracellular protein-tyrosine kinase activity. As a result, autophosphorylation of tyrosine residues in the C-terminal domain of EGFR occurs, which results in activation of several downstream signaling cascades, including the Ras-RAF-MEK-ERK, PI3 kinase-Akt, PLCγ-PKC, and STAT. EGFR phosphorylation at serine 695 is reported to be only partial and occurs only if threonine 693 is phosphorylated. EGFR also undergoes mono and polyubiquitination upon EGF stimulation. However, this ubiquitination does not affect tyrosine kinase activity of the receptor or its signaling capacity. EGFR is commonly overexpressed and is mutated in many human malignancies and is often associated with aggressive phenotypes.
Reacts with external domain of EGF receptor on all human cells. Competes with EGF and TGF-α for binding on human cells.

Specificity

Recognizes both phosphorylated and non-phosphorylated human EGF receptors.

Immunogen

Human A431 membrane proteins.

Application

Detect EGFR using this Anti-EGFR Antibody, neutralizing, clone LA1 validated for use in Immuncytochemistry, Immunohistochemistry, Immunoprecipitation, Neutralization, and Western Blotting.

Immunocytochemistry (IC) Analysis: 2μg/mL of a representative lot detected EGFR in A431 cells.
Immunohistochemistry (Paraffin) (IHC) Analysis: A 1:50 dilution of this antibody detected EGFR in Human placenta tissue sections.
Immunoprecipitation Analysis: A representative lot of this antibody immunoprecipitated EGFR in Huh-7.5 cells. (Diao, J., et al. (2012). J. Virol. 86(20); 10935-10949).
Western Blotting Analysis: 1 mg/mL of a representative lot detected EGFR in A431 cell lysate.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

routinely evaluated on RIPA lysates from non-stimulated human A431 cells

Target description

180 kDa

Linkage

Replaces: 04-337; 04-338

Physical form

Format: Purified
Lyophilized protein G Purified mouse immunoglobulin. Lyophilized from 0.1M Tris-glycine, pH 7.4, 0.15M NaCl containing no preservatives.
Protein G Purified

Storage and Stability

Lyophilized: 2 years at -20°C; Rehydrated: 1 month at 4°C or 6 months at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Autocrine regulation of DU145 human prostate cancer cell growth by epidermal growth factor-related polypeptides.
J M Connolly, D P Rose
Prostate null
Catherine Le Quément et al.
American journal of physiology. Lung cellular and molecular physiology, 294(6), L1076-L1084 (2008-04-09)
Macrophage metalloelastase (MMP-12) is described to be involved in pulmonary inflammatory response. To determine the mechanisms linking MMP-12 and inflammation, we examined the effect of recombinant human MMP-12 (rhMMP-12) catalytic domain on IL-8/CXCL8 production in cultured human airway epithelial (A549)
Manabu Chokki et al.
American journal of respiratory cell and molecular biology, 30(4), 470-478 (2003-09-23)
Human airway trypsin-like protease (HAT) is a serine protease found in sputum of patients with chronic airway diseases and is an agonist of protease-activated receptor-2 (PAR-2). Results from this study show that HAT treatment also enhances mucus production by the
Christina L Hirota et al.
American journal of physiology. Gastrointestinal and liver physiology, 303(1), G111-G119 (2012-04-21)
Proteinase-activated receptor (PAR)(2), a G protein-coupled receptor activated by serine proteinases, has been implicated in both intestinal inflammation and epithelial proliferation. Cyclooxygenase (COX)-2 is overexpressed in the gut during inflammation as well as in colon cancer. We hypothesized that PAR(2)
Yan Guan et al.
Science advances, 1(9), e1500633-e1500633 (2015-11-26)
Measuring small molecule interactions with membrane proteins in single cells is critical for understanding many cellular processes and for screening drugs. However, developing such a capability has been a difficult challenge. We show that molecular interactions with membrane proteins induce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service