Skip to Content
Merck
All Photos(1)

Documents

CS0030

Sigma-Aldrich

Senescence Cells Histochemical Staining Kit

sufficient for 100 tests

Synonym(s):

Senescent cells IHC kit, cellular β-galactosidase probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.32

usage

sufficient for 100 tests

Quality Level

packaging

pkg of 1 kit

storage condition

dry at room temperature

detection method

colorimetric

shipped in

dry ice

storage temp.

−20°C

General description

The Senescence Cells Histochemical Staining Kit contains all the reagents required for identifying senescent cells using an assay based on a histochemical stain for ß-galactosidase activity at pH 6. ß-galactosidase activity is detectable in senescent cells, but not in quiescent, immortal, or tumor cells.
The Senescence Cells Histochemical Staining Kit contains all the reagents required for identifying senescent cells using an assay based on a histochemical stain for β-galactosidase activity at pH 6. β-galactosidase activity is detectable in senescent cells, but not in quiescent, immortal, or tumor cells. Cellular senescence is a progression of events, whereby cells move from an actively dividing to a non-dividing stage. The cellular senescence process is associated with aging. The decrease in cell division is virtually irreversible and complete. In conjunction with the loss of the ability to divide, changes occur in the morphology, shape, and physical appearance of the cells, and their pattern of gene expression. At the end of the process cell death usually occurs, although the cells may remain viable for a long time.

Application

Senescence Cells Histochemical Staining Kit has been used for:
  • senescence-associated β-galactosidase assay
  • to test for senescence in human pancreatic cancer cells
  • adipose-derived stem cells (ADSCs)
  • cord blood mesenchymal stromal/stem cells (CBMSC)

Biochem/physiol Actions

Replicative senescence is a growth-arrest state associated with loss of division potential, changes in cell morphology, shape and physical appearance, and the pattern of gene expression in cells.

Legal Information

Manufactured under license to US Patent Nos. 5,491,069 and 5,795,728.

Kit Components Only

Product No.
Description

  • X-gal solution 4 mL

  • Staining Solution, 10X 15 mL

  • Fixation Buffer 10x 15 mL

  • Reagent B 1.5 mL

  • Reagent C 1.5 mL

  • Dulbecco's Phosphate Buffered Saline (PBS) 10x 60 mL

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mandy Oj Grootaert et al.
Autophagy, 11(11), 2014-2032 (2015-09-24)
Autophagy is triggered in vascular smooth muscle cells (VSMCs) of diseased arterial vessels. However, the role of VSMC autophagy in cardiovascular disease is poorly understood. Therefore, we investigated the effect of defective autophagy on VSMC survival and phenotype and its
Natalja E Fedorovich et al.
Biomaterials, 30(3), 344-353 (2008-10-22)
Photopolymerizable hydrogels, formed by UV-exposure of photosensitive polymers in the presence of photoinitiators, are widely used materials in tissue engineering research employed for cellular entrapment and patterning. During photopolymerization, the entrapped cells are directly exposed to polymer and photoinitiator molecules.
B van der Loo et al.
Experimental cell research, 241(2), 309-315 (1998-06-25)
A beta-galactosidase activity has recently been used as a histochemical marker of replicative senescence in human fibroblasts and keratinocytes. To establish whether this marker could be used to detect senescence of vascular cells, we have investigated its presence in cultures
G P Dimri et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(20), 9363-9367 (1995-09-26)
Normal somatic cells invariably enter a state of irreversibly arrested growth and altered function after a finite number of divisions. This process, termed replicative senescence, is thought to be a tumor-suppressive mechanism and an underlying cause of aging. There is
Mafalda Ramos de Matos et al.
Cancers, 11(3) (2019-03-23)
Intratumor genetic heterogeneity (ITH) is the main obstacle to effective cancer treatment and a major mechanism of drug resistance. It results from the continuous evolution of different clones of a tumor over time. However, the molecular features underlying the emergence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service