Skip to Content
Merck
All Photos(1)

Key Documents

P-045

Supelco

Pheniramine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2
CAS Number:
Molecular Weight:
240.34
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CN(C)CCC(c1ccccc1)c2ccccn2

InChI

1S/C16H20N2/c1-18(2)13-11-15(14-8-4-3-5-9-14)16-10-6-7-12-17-16/h3-10,12,15H,11,13H2,1-2H3

InChI key

IJHNSHDBIRRJRN-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

General description

Pheniramine is an antihistamine with sedative effects used for the treatment of hay fever. This certified solution standard is suitable as starting material for the preparation of calibrators and controls in pheniramine testing by GC/MS or LC-MS/MS. Pheniramine is found in over-the-counter cold medicines and in eye-drops such as Visine brand eye drops.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ho Jun Lee et al.
Journal of Korean medical science, 24(5), 879-882 (2009-10-02)
The purpose of this study was to determine the effectiveness of antihistamine therapy for withdrawal movements caused by rocuronium injection. One hundred seventy one ASA I-II adults undergoing elective surgery were randomly assigned to one of two groups. Patients in
Ching-Hsia Hung et al.
European journal of pharmacology, 673(1-3), 20-24 (2011-11-03)
The aim of this study was to evaluate the local anesthetic effects of pheniramine and diphenhydramine, two histamine H₁ receptor antagonists, on spinal anesthesia and their comparison with lidocaine, a commonly used local anesthetic. After rats were injected intrathecally with
Tshering Lama et al.
Journal of telemedicine and telecare, 17(2), 105-108 (2010-12-09)
We describe a young woman from a rural village in Nepal who suffered multiple wasp and hornet stings. She collapsed and was managed by a telephone consultation between a village health worker and a hospital specialist. The patient recovered fully.
Ana B Guimaraes-Correa et al.
Viruses, 3(7), 1041-1058 (2011-10-14)
Adult T-cell leukemia/lymphoma (ATLL) is a highly aggressive disease that occurs in individuals infected with the human T lymphotropic virus type 1 (HTLV-1). Patients with aggressive ATLL have a poor prognosis because the leukemic cells are resistant to conventional chemotherapy.
Kerem Karaman et al.
American journal of surgery, 205(2), 213-219 (2012-09-05)
Laparoscopic cholecystectomy (LC) still leads to significant postoperative nausea and vomiting (PONV) and pain. Our aim was to evaluate the efficacy of dexamethasone or pheniramine hydrogen maleate, either alone or combined, in reducing the stress response and symptoms after LC.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service