Skip to Content
Merck
All Photos(1)

Key Documents

N-042

Supelco

Naproxen solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14O3
CAS Number:
Molecular Weight:
230.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

COc1ccc2cc(ccc2c1)[C@H](C)C(O)=O

InChI

1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m0/s1

InChI key

CMWTZPSULFXXJA-VIFPVBQESA-N

Gene Information

General description

Naproxen is a nonsteroidal anti-inflammatory drug (NSAID) sold under the trade name Aleve®. This COX inhibitor is indicated for the treatment of pain, fever, inflammation and stiffness. This Certified Spiking Solution® is applicable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or pharmaceutical research.

Legal Information

Aleve is a registered trademark of Bayer Healthcare LLC
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenny-Maria Brozinski et al.
Environmental science & technology, 47(1), 342-348 (2012-11-29)
Pharmaceutical residues are ubiquitous in rivers, lakes, and at coastal waters affected by discharges from municipal wastewater treatment plants. In this study, the presence of 17 different pharmaceuticals and six different phase I metabolites was determined in the bile of
Christopher Lockhart et al.
Biophysical journal, 103(11), 2341-2351 (2013-01-04)
All-atom explicit solvent model and replica exchange molecular dynamics were used to investigate binding of Alzheimer's biomarker FDDNP to the Aβ(10-40) monomer. At low and high concentrations, FDDNP binds with high affinity to two sites in the Aβ(10-40) monomer located
A Li-Wan-Po et al.
Journal of clinical pharmacy and therapeutics, 38(1), 36-40 (2012-11-22)
In analgesic trials, pain relief is often assessed using a pain-relief score. We aimed to assess, through a meta-analysis, whether absence of need for rescue medication (NNR) is a reliable outcome measure in the evaluation of acute pain relief. Individual-patient
Mei Zhang et al.
Antimicrobial agents and chemotherapy, 57(1), 484-489 (2012-11-14)
A rapid and simple high-performance liquid chromatography (HPLC) assay was developed for the simultaneous determination of three triazole antifungals (voriconazole, posaconazole, and itraconazole and the metabolite of itraconazole, hydroxyitraconazole) in human plasma. Sample preparation involved a simple one-step protein precipitation
Raffaele Marotta et al.
Water research, 47(1), 373-383 (2012-11-06)
The kinetics of photodegradation of the non steroidal anti-inflammatory drug naproxen (+)-S-2-(6-methoxynaphthalen-2-yl)propanoic acid, an emerging organic pollutant, was studied in aqueous solutions under deaerated and aerated conditions. The photolysis experiments were carried out under monochromatic irradiation (λ = 254 nm)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service