Skip to Content
Merck
All Photos(1)

Documents

P-906

Supelco

Prazepam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17ClN2O
CAS Number:
Molecular Weight:
324.80
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1ccc2N(CC3CC3)C(=O)CN=C(c4ccccc4)c2c1

InChI

1S/C19H17ClN2O/c20-15-8-9-17-16(10-15)19(14-4-2-1-3-5-14)21-11-18(23)22(17)12-13-6-7-13/h1-5,8-10,13H,6-7,11-12H2

InChI key

MWQCHHACWWAQLJ-UHFFFAOYSA-N

General description

Prazepam is a benzodiazepine prescribed for the short-term treatment of anxiety. Sold under the trade name Centrax, this drug also has anticonvulsant, sedative, and muscle relaxant effects. This Snap-N-Spike® Reference Solution is suitable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paraskevi Valavani et al.
Journal of mass spectrometry : JMS, 40(4), 516-526 (2005-02-16)
A method was developed and fully validated for the quantitation of prazepam and its major metabolites, oxazepam and nordiazepam, in human plasma. Sample pretreatment was achieved by solid-phase extraction using Oasis HLB cartridges. The extracts were analysed by high-performance liquid
Polyuria after olanzapine overdose.
Leon Etienne et al.
The American journal of psychiatry, 161(6), 1130-1130 (2004-06-01)
W D Hooper et al.
Biochemical pharmacology, 43(6), 1377-1380 (1992-03-17)
Phenobarbitone-pretreated male Sprague-Dawley rat liver microsomes were used to examine C3-hydroxylation and N-dealkylation of four clinically important benzodiazepines: diazepam (DZP), prazepam (PZP), pinazepam (PIN) and halazepam (HZP). These substrates differ only in the nature of the N-substituent of the B
M Ansseau et al.
L'Encephale, 17(4), 291-294 (1991-07-01)
In order to assess the clinical usefulness of benzodiazepine brief therapy with planned tapering, prazepam as drops was administered to 40 psychiatric outpatients suffering from generalized anxiety disorder. After a one-week placebo period, the patients received prazepam 40 mg daily
P Lemoine et al.
Pharmacopsychiatry, 24(5), 175-179 (1991-09-01)
The efficacy of antidepressants is well established in major depressions, especially those with melancholic features. However, some anxiolytics also appear to have antidepressant properties at least for outpatients. 118 outpatients (25 males, 93 females, age: 18-60) with major depression according

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service