Skip to Content
Merck
All Photos(1)

Key Documents

M-007

Supelco

(±)-Methadone solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27NO
CAS Number:
Molecular Weight:
309.45
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCC(C(CC(C)N(C)C)(C1=CC=CC=C1)C2=CC=CC=C2)=O

InChI

1S/C21H27NO/c1-5-20(23)21(16-17(2)22(3)4,18-12-8-6-9-13-18)19-14-10-7-11-15-19/h6-15,17H,5,16H2,1-4H3

InChI key

USSIQXCVUWKGNF-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

Looking for similar products? Visit Product Comparison Guide

General description

Methadone is a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. The drug is sold under several trade names such as Symoron, Dolophine®, Amidone, Methadose®, Physeptone, or Heptadon. This Certified Spiking Solution® is suitable for LC/MS or GC/MS testing methods in pain prescription monitoring, forensic analysis or clinical toxicology.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Dolophine is a registered trademark of Roxane Laboratories, Inc.
Methadose is a registered trademark of Mallinckrodt Inc.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maintenance medication for opiate addiction: the foundation of recovery.
Bart G.
Journal of Addictive Diseases, 31(3), 207-225 (2012)
Advice on methadone and oral health: dental public health in action.
C M Jones et al.
Community dental health, 30(2), 68-70 (2013-07-31)
Kobra Lashkaripour et al.
JPMA. The Journal of the Pakistan Medical Association, 62(10), 1003-1007 (2013-07-23)
To examine the 3-month effect of methadone maintenance treatment (MMT) on quality of life (QOL) of substance-dependent outpatients in Iran. The qausi-experimental study performed at the Oral Maintenance Methadone Treatment Clinic of the Baharan Psychiatric Hospital, a university hospital in
David Kao et al.
Annals of internal medicine, 158(10), 735-740 (2013-05-22)
Long-acting opioids are a leading cause of accidental death in the United States, and methadone is associated with greater mortality rates. Whether this increase is related to the proarrhythmic properties of methadone is unclear. To describe methadone-associated arrhythmia events reported
The lethal burden of drug overdose.
Lancet (London, England), 382(9895), 833-833 (2013-09-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service