Skip to Content
Merck
All Photos(1)

Key Documents

N-071

Supelco

(±)-Nornicotine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2
CAS Number:
Molecular Weight:
148.20
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

C1CNC(C1)c2cccnc2

InChI

1S/C9H12N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1,3,5,7,9,11H,2,4,6H2

InChI key

MYKUKUCHPMASKF-UHFFFAOYSA-N

General description

Nornicotine is a tobacco alkaloid and nicotine metabolite. Since virtually all tobacco products contain nornicotine, smoking or chewing tobacco exposes the user to this nicotine alkaloid. In urine testing by GC/MS or LC/MS, nornicotine is used as an indicator of smoking or use of chewing tobacco or nicotine replacement products.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph R Holtman et al.
Pharmacology, biochemistry, and behavior, 94(3), 352-362 (2009-10-06)
Neuronal nicotinic acetylcholinic receptors (nAChR) are promising targets for the development of novel analgesics. Nicotine and other nAChR-agonists produce profound analgesia in rodent models of acute and persistent pain. However, significant side-effects are of concern. Nornicotine (N-desmethyl-nicotine) appears to activate
Wojciech Piekoszewski et al.
Przeglad lekarski, 66(10), 593-597 (2009-01-01)
The assay of biomarkers in biological material is the most popular and reliable method in estimate exposure to tobacco smoke. Nicotine and its metabolites qualify to the most specific biomarkers for tobacco smoke. Currently the most often used are cotinine
Piotr Tutka et al.
Nicotine & tobacco research : official journal of the Society for Research on Nicotine and Tobacco, 10(8), 1385-1390 (2008-08-08)
Smoking during pregnancy remains a major public health concern and is associated with numerous adverse effects. Recently the clearance of nicotine and cotinine was shown to be substantially increased in pregnant women compared with nonpregnant controls. The present study investigated
François Marclay et al.
Forensic science international, 213(1-3), 73-84 (2011-07-02)
Tobacco consumption is a global epidemic responsible for a vast burden of disease. With pharmacological properties sought-after by consumers and responsible for addiction issues, nicotine is the main reason of this phenomenon. Accordingly, smokeless tobacco products are of growing popularity
Snehal Jadey et al.
The Journal of general physiology, 141(1), 95-104 (2013-01-02)
A primary target for nicotine is the acetylcholine receptor channel (AChR). Some of the ability of nicotine to activate differentially AChR subtypes has been traced to a transmitter-binding site amino acid that is glycine in lower affinity and lysine in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service