Skip to Content
Merck
All Photos(1)

Documents

H-107

Supelco

1,25-Dihydroxyvitamin D3-13C3 (25,26,27-13C3) solution

5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Deuterated 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C24H44O3
Molecular Weight:
419.61
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

5 μg/mL in ethanol

mp

-143.99 °C

application(s)

clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

O[C@@H](C[C@H](O)C/1=C)CC1=C\C=C2[C@@](CC[C@]3([H])[C@H](C)CCC[13C](O)([13CH3])[13CH3])([H])[C@]3(C)CCC\2

InChI

1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23-,24+,25+,27-/m1/s1/i3+1,4+1,26+1

InChI key

GMRQFYUYWCNGIN-SYUIRNBGSA-N

General description

A stable-labeled internal standard suitable for quantitation of 1,25-Dihydroxyvitamin D3 levels by LC-MS/MS. 1,25-Dihydroxyvitamin D3, also known as calcitriol, is a hormonally active metabolite of vitamin D responsible for regulating calcium. Measurement of 1,25-Dihydroxyvitamin D3 is used to distinguish some cases of primary hyperparathyroidism from hypercalcemia of cancer and for the differential diagnosis of vitamin D-dependent rickets.

Application


  • Leigh Syndrome with HIBCH Mutations: Discusses the metabolic effects of mutations related to Leigh syndrome and includes information on 3-hydroxybutyryl carnitine isomers (Wang et al., 2021, frontiersin.org).

  • Highly Efficient Ketone Body Treatment in Leukodystrophy: Highlights the therapeutic potential of L-carnitine in treatments, specifically focusing on the metabolism of L-3-hydroxybutyryl-CoA in the context of fatty acid β-oxidation disorders (Gautschi et al., 2015, nature.com).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark A Crumling et al.
PloS one, 7(12), e53280-e53280 (2013-01-04)
Cyclodextrins are sugar compounds that are increasingly finding medicinal uses due to their ability to complex with hydrophobic molecules. One cyclodextrin in particular, 2-hydroxypropyl-β-cyclodextrin (HPβCD), is used as a carrier to solubilize lipophilic drugs and is itself being considered as
Fang Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(26), 12986-12995 (2019-06-12)
The aberrant hedgehog (Hh) pathway plays important roles in multiple cancer types, therefore serving as a promising drug target. Current clinically available hedgehog-targeted drugs act mostly by antagonizing the upstream component smoothened; however, both primary and acquired resistance to FDA-approved
Eric Wang et al.
Cancer cell, 35(3), 369-384 (2019-02-26)
RNA-binding proteins (RBPs) are essential modulators of transcription and translation frequently dysregulated in cancer. We systematically interrogated RBP dependencies in human cancers using a comprehensive CRISPR/Cas9 domain-focused screen targeting RNA-binding domains of 490 classical RBPs. This uncovered a network of
Kornél Király et al.
Scientific reports, 8(1), 3490-3490 (2018-02-24)
Altered pain sensations such as hyperalgesia and allodynia are characteristic features of various pain states, and remain difficult to treat. We have shown previously that spinal application of dipeptidyl peptidase 4 (DPP4) inhibitors induces strong antihyperalgesic effect during inflammatory pain.
Matthew B Pomrenze et al.
Cell reports, 29(1), 13-21 (2019-10-03)
Central amygdala (CeA) neurons that produce corticotropin-releasing factor (CRF) regulate anxiety and fear learning. These CeACRF neurons release GABA and several neuropeptides predicted to play important yet opposing roles in these behaviors. We dissected the relative roles of GABA, CRF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service