Skip to Content
Merck
All Photos(1)

Documents

17937

Sigma-Aldrich

25-Hydroxyvitamin D2

≥95.0% (HPLC)

Synonym(s):

25-Hydroxycalciferol, 25-Hydroxyergocalciferol, Ercalcidiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O2
CAS Number:
Molecular Weight:
412.65
Beilstein:
4716773
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058017
PubChem Substance ID:
NACRES:
NA.28

biological source

synthetic

Assay

≥95.0% (HPLC)

form

crystalline powder

color

white to off-white

mp

88.0-95.0 °C

storage temp.

−70°C

SMILES string

[H][C@@]12CC[C@H]([C@H](C)\C=C\[C@H](C)C(C)(C)O)[C@@]1(C)CCC/C2=C\C=C3\C[C@@H](O)CCC3=C

InChI

1S/C28H44O2/c1-19-10-14-24(29)18-23(19)13-12-22-8-7-17-28(6)25(15-16-26(22)28)20(2)9-11-21(3)27(4,5)30/h9,11-13,20-21,24-26,29-30H,1,7-8,10,14-18H2,2-6H3/b11-9+,22-12+,23-13-/t20-,21+,24+,25-,26+,28-/m1/s1

InChI key

KJKIIUAXZGLUND-ICCVIKJNSA-N

Biochem/physiol Actions

Ergocalciferol (vitamin D2) is activated in vivo by hydroxylation to 25-hydroxyergocalciferol and 1α,25-dihydroxycalciferol. Activated Vitamin D2 molecules may be used in a wide range of studies to assess their effects on functions such as immune function and calcium homeostasis.

Packaging

Amber polypropylene vial

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin D Cashman et al.
The British journal of nutrition, 112(2), 193-202 (2014-05-02)
It has been suggested that vitamin D₂ is not very prevalent in the human food chain. However, data from a number of recent intervention studies suggest that the majority of subjects had measurable serum 25-hydroxyvitamin D₂ (25(OH)D₂) concentrations. Serum 25(OH)D₂
Paul Haggarty et al.
The British journal of nutrition, 109(5), 898-905 (2012-11-22)
The aims of the present study were to determine compliance with current advice on vitamin D and to assess the influence of season, dietary intake, supplement use and deprivation on vitamin D status in pregnant mothers and newborns in the
Grant R Campbell et al.
PLoS pathogens, 8(11), e1003017-e1003017 (2012-11-21)
Toll-like receptors (TLR) are important in recognizing microbial pathogens and triggering host innate immune responses, including autophagy, and in the mediation of immune activation during human immunodeficiency virus type-1 (HIV) infection. We report here that TLR8 activation in human macrophages
Amin Salehpour et al.
Nutrition journal, 11, 78-78 (2012-09-25)
Vitamin D concentrations are linked to body composition indices, particularly body fat mass. Relationships between hypovitaminosis D and obesity, described by both BMI and waist circumference, have been mentioned. We have investigated the effect of a 12-week vitamin D3 supplementation
Stephen J Bruce et al.
Rapid communications in mass spectrometry : RCM, 27(1), 200-206 (2012-12-15)
The aim of the work was to develop and validate a method for the quantification of vitamin D metabolites in serum using ultra-high-pressure liquid chromatography coupled to mass spectrometry (LC/MS), and to validate a high-resolution mass spectrometry (LC/HRMS) approach against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service