Skip to Content
Merck
All Photos(1)

Documents

H-083

Supelco

25-Hydroxyvitamin D3 solution

100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in ethanol

application(s)

clinical testing
clinical testing
clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

O.C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2C(\CCC[C@]12C)=C\C=C3\C[C@@H](O)CCC3=C

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23+,24-,25+,27-/m1/s1

InChI key

JWUBBDSIWDLEOM-DTOXIADCSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® of 25-Hydroxyvitamin D3, a prohormone whose blood serum levels are used to determine Vitamin D nutritional status. This solution standard is suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls and calibrators for LC/UV and LC-MS/MS assays; as system tuning/suitability standards; linearity verification; internal proficiency testing; and for OEM use in preparation of reagents, development of assays and instrument IQ/OQ. The stable, quantitative solution format provides significant benefits to the user including material savings, increased efficiency of labor, and convenience of use. The shelf stable Snap-N-Spike® format allows laboratories to eliminate painstaking weighing operations and provides the ability to spike a known concentration of analyte into their matrix of choice just before use.

Application

25-Hydroxyvitamin D3 may be used as a certified reference material (CRM) for the determination of the analyte in biological samples by chromatography techniques.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Soares et al.
Poultry science, 74(12), 1919-1934 (1995-12-01)
Vitamin D is a complex of secosteroids that must undergo metabolic alterations to reach optimal biological activity. The parent compounds 1) ergocalciferol (D2) and 2) cholecalciferol (D3) can be synthesized in the leaves of many plants or in the skin
Development and validation of an LC--MS/MS based method for quantification of 25 hydroxyvitamin D2 and 25 hydroxyvitamin D3 in human serum and plasma
Zhang SW, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 961(1), 62-70 (2014)
Antrison Morris et al.
Poultry science, 93(8), 1951-1956 (2014-06-17)
Three experiments were conducted to study the effects of 25-hydroxycholecalciferol supplementation on BW gain, IL-1β, and 1α-hydroxylase mRNA expression in different organs of broiler chickens following a lipopolysaccharide (LPS) injection. In experiment I, birds were fed a basal diet supplemented
A method for simultaneous determination of 25-hydroxyvitamin D2 and 25-hydroxyvitamin D3 in human plasma by using two steps of high-performance liquid chromatography
Okano T, et al.
J. Nutr. Sci. Vitaminol., 27(1), 43-54 (1981)
Maria Samefors et al.
European journal of endocrinology, 170(5), 667-675 (2014-02-13)
Institutionalised elderly people at northern latitudes may be at elevated risk for vitamin D deficiency. In addition to osteoporosis-related disorders, vitamin D deficiency may influence several medical conditions conferring an increased mortality risk. The aim of this study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service