Skip to Content
Merck
All Photos(1)

Documents

M-035

Supelco

Meperidine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO2
CAS Number:
Molecular Weight:
247.33
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

InChI

1S/C15H21NO2/c1-3-18-14(17)15(9-11-16(2)12-10-15)13-7-5-4-6-8-13/h4-8H,3,9-12H2,1-2H3

InChI key

XADCESSVHJOZHK-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

General description

Meperidine is an opioid analgesic commonly known by its brand name, Demerol®. This certified solution standard is applicable for use in urine drug testing, pain prescription monitoring, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS. This painkiller, considered to have a fast onset of action, has a history of recreational abuse.

Application


  • Rapid and Convenient Potentiometric Method for Determining Fluorosulfate, a Byproduct of the Fumigant and Greenhouse Gas Sulfuryl Fluoride.: This study presents a potentiometric method using silver sulfate for detecting fluorosulfate, a byproduct of sulfuryl fluoride, highlighting its utility in environmental monitoring and analytical chemistry (Chen et al., 2024).

  • Biosynthesis of Ag-Pd bimetallic alloy nanoparticles through hydrolysis of cellulose triggered by silver sulfate.: This research demonstrates the biosynthesis of silver-palladium alloy nanoparticles using silver sulfate, offering insights into green synthesis approaches and applications in catalysis and material science (Li et al., 2018).

  • Self-Assembled Metal-Organic Biohybrids (MOBs) Using Copper and Silver for Cell Studies.: The paper explores the creation of biohybrid structures using silver sulfate, showing potential applications in cellular biology and bioanalytical chemistry (Karekar et al., 2019).

  • Modified APHA closed-tube reflux colorimetric method for TOC determination in water and wastewater.: The study introduces a modified method using silver sulfate for determining total organic carbon in water and wastewater, emphasizing advancements in analytical methods for environmental monitoring (Salihu and Bakar, 2018).



Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Demerol is a registered trademark of Sanofi-Aventis
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R M Vassiliadis et al.
Anaesthesia and intensive care, 41(1), 113-115 (2013-02-01)
Pethidine (meperidine) is a unique member of the opioid family. In addition to its analgesic activity, it also has significant local anaesthetic activity. This property enables it to be used as the sole agent for spinal anaesthesia. We describe the
Christoph Marcus Bucher et al.
Journal of clinical pathology, 66(1), 24-28 (2012-10-30)
The diagnostic and clinical usefulness of a powered bone marrow biopsy device (OnControl()) versus a standard manual device (TRAP Hospital System) was studied. Primary endpoints were biopsy quality and patient pain during the procedure. Fifty patients underwent a total of
Alda L Bourque et al.
Gastroenterology nursing : the official journal of the Society of Gastroenterology Nurses and Associates, 35(5), 308-312 (2012-09-29)
The purpose of this study was to determine whether the use of Reiki decreases the amount of meperidine administered to patients undergoing screening colonoscopy. The literature review reveals limited studies to show whether Reiki has been able to decrease the
Hsiao-Wen Tsai et al.
Obstetrics and gynecology, 121(3), 526-531 (2013-05-03)
To estimate the effectiveness of combined intervention with the pulmonary recruitment maneuver and intraperitoneal normal saline infusion to reduce postlaparoscopic shoulder and upper abdominal pain. Patients were randomly assigned to undergo the combined intervention (n=50) or to a control group
Jin Gu Kang et al.
Journal of clinical anesthesia, 24(6), 465-470 (2012-09-19)
To evaluate whether intraoperative low-dose lidocaine infusion decreases postoperative analgesic consumption, ileus, and duration of hospital stay. Prospective, randomized, double-blinded trial. Operating room in a university hospital. 48 ASA physical status 1 and 2 men scheduled for subtotal gastrectomy. Patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service