Skip to Content
Merck
All Photos(1)

Key Documents

E-081

Supelco

Etizolam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClN4S
CAS Number:
Molecular Weight:
342.85
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CCC1=CC2=C(S1)N3C(CN=C2C4=CC=CC=C4Cl)=NN=C3C

InChI

1S/C17H15ClN4S/c1-3-11-8-13-16(12-6-4-5-7-14(12)18)19-9-15-21-20-10(2)22(15)17(13)23-11/h4-8H,3,9H2,1-2H3

InChI key

VMZUTJCNQWMAGF-UHFFFAOYSA-N

General description

A certified solution standard for use in LC/MS or GC/MS benzodiazepine testing methods in clinical toxicology, urine drug testing, or forensic analysis. Etizolam, sold under brand names such as Etilaam, Etizola, or Depas, is a benzodiazepine analog used for the short-term treatment of insomnia, anxiety, or panic attacks. Etizolam is also sold on the designer drug market with cases of abuse noted in Europe and the US.

Application

  • A strategy for the detection of benzodiazepine drugs using low-resolution paper-spray mass spectrometry for harm reduction drug checking: This study explores the application of Etizolam in the development of low-cost, rapid mass spectrometric techniques for detecting benzodiazepines at harm reduction sites, emphasizing its utility in public health interventions (Miskulin et al., 2023).
  • Unexpected results found in larvae samples from two postmortem forensic cases: Incorporates Etizolam to elucidate drug metabolism in forensic toxicology, highlighting its relevance in the postmortem analysis and its impact on forensic investigations (Groth et al., 2022).
  • Rapid and accurate etizolam detection using surface-enhanced Raman spectroscopy for community drug checking: Demonstrates the use of Etizolam as a standard in developing cutting-edge, reliable spectroscopic methods for quick drug testing in community settings, thus advancing forensic and clinical toxicology (Gozdzialski et al., 2022).
  • Incorporation of five common hypnotics into hair after a single dose and application to a forensic case of drug facilitated crimes: This research utilizes Etizolam in studies focused on drug deposition in hair, providing crucial insights for the forensic analysis of drug-facilitated crimes (Sasaki et al., 2021).

Biochem/physiol Actions

Etizolam is a thienodiazepine anxiolytic and as compared to other benzodiazepines has lower dependence potential. It differs from other benzodiazepines in that its molecules possess a thiophene ring instead of benzene. It has anticonvulsant, sedative, anxiolytic, amnesic, hypnotic and muscle relaxant properties. It′s mode of action is through benzodiazepine receptor. It is used to treat insomnia, moderate depression and social anxiety disorder, and has occasionally been associated with blepharospasm and skin lesions.[1][2]

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minoru Narita et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 31(4), 739-750 (2005-08-27)
Clinically, it has been reported that chronic pain induces depression, anxiety, and reduced quality of life. The endogenous opioid system has been implicated in nociception, anxiety, and stress. The present study was undertaken to investigate whether chronic pain could induce
Akira Namera et al.
Chudoku kenkyu : Chudoku Kenkyukai jun kikanshi = The Japanese journal of toxicology, 24(1), 27-34 (2011-04-13)
The immunoassay screening of benzodiazepines in urine is one of the most important methods of drug analysis in clinical and forensic laboratories. We experienced an unusual case of poisoning wherein the result of Triage DOA immunoassay screening was negative, although
Takeshi Terao et al.
Progress in neuro-psychopharmacology & biological psychiatry, 31(1), 295-296 (2006-08-19)
A 65-year-old female patient with major depressive disorder suffered from clonus, shivering and impaired visual acuity after 20 mg/day of paroxetine administration. The symptoms were initially regarded as further manifestations of her somatic symptoms of depression, and paroxetine was increased
Naoko Tanaka et al.
Soudni lekarstvi, 56(3), 38-39 (2011-09-06)
A fatal poisoning case involving etizolam, phenobarbital, promethazine and chlorpromazine is presented. Quantitative toxicological analysis showed that the concentrations of etizolam, phenobarbital, promethazine and chlorpromazine in the femoral blood were 86 ng/ml, 5082 microg/ml, 0.107 microg/ml and 0.144 microg/ml, respectively
Maria Pia De Candia et al.
Clinical therapeutics, 31(12), 2851-2859 (2010-01-30)
Etizolam is an anxiolytic drug with a pharmacologic profile similar to that of the classic benzodiazepines. Neurochemical research suggests that etizolam may have selectivity for the subpopulation of Y-aminobutyric acid type A receptors associated with anxiety (ie, alpha1, beta2, gamma2).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service