Skip to Content
Merck
All Photos(1)

Key Documents

H-129

Supelco

α-Hydroxyetizolam solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClN4OS
CAS Number:
Molecular Weight:
358.85
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[s]1c2c(cc1C(O)C)C=NC(c4[n]2c(nn4)C)c3c(cccc3)Cl

InChI

1S/C17H15ClN4OS/c1-9(23)14-7-11-8-19-15(12-5-3-4-6-13(12)18)16-21-20-10(2)22(16)17(11)24-14/h3-9,15,23H,1-2H3

InChI key

BTGCWXDWJAGZML-UHFFFAOYSA-N

General description

A major urinary metabolite of the benzodiazepine etizolam. Etizolam, commonly sold under the trade name Etilaam, Etizola, or Depas, is a benzodiazepine analog used for the short-term treatment of insomnia, anxiety, or panic attacks. This certified Snap-N-Spike®; solution standard is suitable for use in LC/MS or GC/MS benzodiazepine testing methods in clinical toxicology, urine drug testing, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Smyth et al.
European journal of immunology, 28(12), 4162-4169 (1998-12-23)
Mouse CD8+ CTL reactive with an H-2Db presented 9-mer peptide of the human papilloma virus 16 (HPV-16) protein E749-57 (RAHYNIVTF) were generated from the splenocytes of wild-type C57BL/6 (B6), B6.perforin-deficient, B6.gld or B6.TNF-deficient mice. In short-term (4 h) assays, CTL
M Gunzer et al.
Immunity, 13(3), 323-332 (2000-10-06)
Cognate interactions of naive T cells with antigen-presenting dendritic cells require physical cell-cell contacts leading to signal induction and T cell activation. Using a three-dimensional collagen matrix videomicroscopy model for ovalbumin peptide-specific activation of murine and oxidative mitogenesis of human
Jeffrey A Luross et al.
Arthritis and rheumatism, 46(6), 1671-1682 (2002-07-13)
The receptor-binding B subunit of Escherichia coli heat-labile enterotoxin (EtxB) is a highly stable, nontoxic protein that is capable of modulating immune responses. This study was conducted to determine whether mucosal administration of EtxB can block collagen-induced arthritis (CIA) and
C E Alexander et al.
European journal of immunology, 31(4), 1199-1210 (2001-04-12)
In this study we show an increased incidence of T cell apoptosis in the liver and spleen of mice infected with Leishmania donovani. T cells from L. donovani-infected mice were found to be increasingly susceptible to CD95-mediated apoptosis in vitro
Anthony R Fehr et al.
Journal of virology, 85(2), 663-674 (2010-11-05)
We have previously reported that a newly annotated gene of human cytomegalovirus (HCMV), UL21a, encodes an early viral protein termed pUL21a. Most notably, the virions of a UL21a deletion virus had markedly reduced infectivity, indicating that UL21a is required to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service