Skip to Content
Merck
All Photos(1)

Key Documents

D-056

Supelco

N-Desethylamiodarone-D4 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21I2NO3D4·HCl
CAS Number:
Molecular Weight:
657.74
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Ic1c(c(cc(c1)C(=O)c2c3c([o]c2CCCC)cccc3)I)OC(C(NCC)([2H])[2H])([2H])[2H].Cl

InChI

1S/C23H25I2NO3.ClH/c1-3-5-9-20-21(16-8-6-7-10-19(16)29-20)22(27)15-13-17(24)23(18(25)14-15)28-12-11-26-4-2;/h6-8,10,13-14,26H,3-5,9,11-12H2,1-2H3;1H/i11D2,12D2;

InChI key

OCQPMJVGWGJLLG-ZYMFQSNRSA-N

General description

A Certified Spiking Solution®and stable-labeled internal standard for use in therapeutic drug monitoring analyses by LC/MS. N-Desethylamiodarone is the primary, active metabolite of amiodarone, an antiarrhythmic agent marketed as Pacerone®, Cordarone®, Aratac, or Atlansil. N-Desethylamiodarone and amiodarone are monitored by clinical labs to ensure patients remain within the drug′s therapeutic range.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Cordarone is a registered trademark of Sanofi S.A.
Pacerone is a registered trademark of Upsher-Smith Laboratories, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service