Skip to Content
Merck
All Photos(1)

Documents

999995P

Avanti

Edelfosine

Avanti Research - A Croda Brand 999995P, powder

Synonym(s):

1-O-octadecyl-2-O-methyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H58NO6P
CAS Number:
Molecular Weight:
523.73
UNSPSC Code:
51191904
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (999995P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999995P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC)COCCCCCCCCCCCCCCCCCC)=O

InChI

1S/C10H24NO5P/c1-6-10(14-5)9-16-17(12,13)15-8-7-11(2,3)4/h10H,6-9H2,1-5H3/t10-/m0/s1

InChI key

GVMCXWJIRSIWFJ-JTQLQIEISA-N

Application

Edelfosine or 1-O-octadecyl-2-O-methyl-sn-glycero-3-phosphocholine might be used:
  • as a non-hydrolysable LysoPC (phospholipid) analog for analyzing its ability to block sexual commitment in Plasmodium falciparum
  • in multilamellar vesicle preparation, to study its effect on model membranes
  • in the selection and screening of mutagenized cells, having the ability to inhibit the transport of alkylphosphocholine drugs across the plasma membrane

Biochem/physiol Actions

Edelfosine acts as a precursor for alkyl-lysophospholipids. It possesses apoptotic action against several cancer cells such as prostate, leukemia, brain and lung tumors. It is associated with cellular transport system, signaling transducing systems, cytokine synthesis and lipid metabolism. Edelfosine is known to control intracellular calcium levels. It is not mutagenic and its anti-tumor action requires its incorporation into the cell. Edelfosine is also found to block the replication of human immunodeficiency virus type I (HIV-I).

Packaging

5 mL Clear Glass Sealed Ampule (999995P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessio Ausili et al.
The journal of physical chemistry. B, 112(37), 11643-11654 (2008-08-21)
The effect of edelfosine (1- O-octadecyl-2- O-methyl-rac-glycero-3-phosphocholine or ET-18-OCH3) on model membranes containing 1-palmitoyl-2-oleoyl- sn-glycero-3-phosphocholine/sphingomyelin/cholesterol (POPC/SM/cholesterol) was studied by several physical techniques. The sample POPC/SM (1:1 molar ratio) showed a broad phase transition as seen by DSC, X-ray diffraction, and
Pamela K Hanson et al.
The Journal of biological chemistry, 278(38), 36041-36050 (2003-07-05)
The alkylphosphocholine class of drugs, including edelfosine and miltefosine, has recently shown promise in the treatment of protozoal and fungal diseases, most notably, leishmaniasis. One of the major barriers to successful treatment of these infections is the development of drug
Bruno M Castro et al.
The journal of physical chemistry. B, 117(26), 7929-7940 (2013-06-07)
Edelfosine (1-O-octadecyl-2-O-methyl-sn-glycero-phosphocholine) and miltefosine (hexadecylphosphocholine) are synthetic alkylphospholipids (ALPs) that are reported to selectively accumulate in tumor cell membranes, inducing Fas clustering and activation on lipid rafts, triggering apoptosis. However, the exact mechanism by which these lipids elicit these events
Nicolas M B Brancucci et al.
Cell, 171(7), 1532-1544 (2017-11-14)
Transmission represents a population bottleneck in the Plasmodium life cycle and a key intervention target of ongoing efforts to eradicate malaria. Sexual differentiation is essential for this process, as only sexual parasites, called gametocytes, are infective to the mosquito vector. Gametocyte production
Alessio Ausili et al.
Langmuir : the ACS journal of surfaces and colloids, 34(28), 8333-8346 (2018-06-21)
Edelfosine is an anticancer drug with an asymmetric structure because, being a derivative of glycerol, it possesses two hydrophobic substituents of very different lengths. We showed that edelfosine destabilizes liquid-ordered membranes formed by either 1-palmitoyl-2-oleoyl- sn-glycero-3-phosphocholine, sphingomyelin (SM), and cholesterol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service