Skip to Content
Merck
All Photos(1)

Key Documents

900400P

Avanti

Palmitic acid (15-yne)

Avanti Research - A Croda Brand 900400P, powder

Synonym(s):

15-hexadecynoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28O2
CAS Number:
Molecular Weight:
252.39
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (900400P-1mg)
pkg of 1 × 5 mg (900400P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900400P

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC(CCCCCCCCCCCCCC#C)=O

InChI

1S/C16H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h1H,3-15H2,(H,17,18)

InChI key

PUZGUNYANHPRKM-UHFFFAOYSA-N

Application

Palmitic acid (15-yne) has been used as a component of fatty acid probe stock solution for the optimization of Alk-C16 incorporation onto cellular proteins. It is also suitable for metabolic labeling.

Biochem/physiol Actions

Alkynyl palmitic acid (aPA) is a modified palmitic acid with an ω-terminal alkyne. The terminal alkyne group can be used in a highly specific linking reaction with azide-containing reagents, known as ‘click chemistry′. The click chemistry reactions are rapid, convenient, versatile and regiospecific. They are easy to purify. aPA can be used for isolating palmitoylated proteins.

Packaging

5 mL Amber Glass Screw Cap Vial (900400P-1mg)
5 mL Amber Glass Screw Cap Vial (900400P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher D Hein et al.
Pharmaceutical research, 25(10), 2216-2230 (2008-05-30)
Click chemistry refers to a group of reactions that are fast, simple to use, easy to purify, versatile, regiospecific, and give high product yields. While there are a number of reactions that fulfill the criteria, the Huisgen 1,3-dipolar cycloaddition of
Christoph Thiele et al.
ACS chemical biology, 7(12), 2004-2011 (2012-09-25)
Fatty acids are abundant constituents of all biological systems, and their metabolism is important for normal function at all levels of an organism. Aberrations in fatty acid metabolism are associated with pathological states and have become a focus of current
Keri A Tallman et al.
Journal of lipid research, 54(3), 859-868 (2013-01-12)
Monitoring lipid distribution and metabolism in cells and biological fluids poses many challenges because of the many molecular species and metabolic pathways that exist. This study describes the synthesis and study of molecules that contain an alkyne functional group as
Xinxin Gao et al.
FEBS letters, 585(15), 2501-2506 (2011-07-12)
Palmitoylation of the Wnt and Hedgehog proteins is critical for maintaining their physiological functions. To date, there are no reported studies that characterize the cellular distribution of the palmitoylated forms of these proteins. Here, we describe the subcellular localization of
Alexander L Ticho et al.
The Journal of biological chemistry, 295(14), 4488-4497 (2020-02-20)
The ileal apical sodium-dependent bile acid transporter (ASBT) is crucial for the enterohepatic circulation of bile acids. ASBT function is rapidly regulated by several posttranslational modifications. One reversible posttranslational modification is S-acylation, involving the covalent attachment of fatty acids to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service