Skip to Content
Merck
All Photos(2)

Key Documents

810131P

Avanti

16:0-12:0 NBD PC

Avanti Research - A Croda Brand 810131P, powder

Synonym(s):

1-palmitoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H74N5O11P
CAS Number:
Molecular Weight:
856.04
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810131P-1mg)
pkg of 5 × 1 mg (810131P-5mg)
pkg of 5 × 2 mg (810131P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810131P

lipid type

fluorescent lipids
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholine (PC) is the most abundant phospholipid in eukaryotes and a prime metabolite of glycerolipid metabolism. It is present in the mucosal layer of the colon. In NBD PC, the fluorescent 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) label is linked to phosphatidylcholine.

Application

16:0-12:0 NBD PC may be used:
  • in the microfluidic chip-based technology to demonstrate separation of fluorophore-labeled phospholipids and to monitor enzyme activities of lipid-modifying enzymes
  • as a fluorophore for 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) in fluorescence recovery experiments
  • as a substrate to test scramblase activity

Biochem/physiol Actions

The use of 7-nitrobenz-2-oxa-1,3-diazol-4-yl phosphatidylcholine (NBD PC) as fluorescent probes in lipid membranes is involved in the study of various cellular processes.

Packaging

5 mL Amber Glass Screw Cap Vial (810131P-10mg)
5 mL Amber Glass Screw Cap Vial (810131P-1mg)
5 mL Amber Glass Screw Cap Vial (810131P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulia Y Tyurina et al.
The Journal of biological chemistry, 282(11), 8498-8509 (2007-01-19)
Macrophage recognition of apoptotic cells depends on externalization of phosphatidylserine (PS), which is normally maintained within the cytosolic leaflet of the plasma membrane by aminophospholipid translocase (APLT). APLT is sensitive to redox modifications of its -SH groups. Because activated macrophages
C Scomparin et al.
The European physical journal. E, Soft matter, 28(2), 211-220 (2008-12-23)
The diffusion law of DMPC and DPPC in Supported Lipid Bilayers (SLB), on different substrates, has been investigated in details by Fluorescence Recovery After Patterned Photobleaching (FRAPP). Over micrometer length scales, we demonstrate the validity of a purely Brownian diffusive
Rachel R Kroe et al.
Analytical biochemistry, 390(1), 1-13 (2008-12-24)
Analytical ultracentrifugation is a widely used method for characterizing the solution behavior of macromolecules. However, the two commonly used detectors, absorbance and interference, impose some fundamental restrictions on the concentrations and complexity of the solutions that can be analyzed. The
Ellen C Wrobel et al.
Physical chemistry chemical physics : PCCP, 22(11), 6154-6166 (2020-03-04)
The antibacterial activity of a calixarene derivative, p-tert-butylcalix[6]arene (Calix6), was assessed and was shown not to inhibit the growth of E. coli, S. aureus and B. subtilis bacteria. With the aim of gaining more insights into the absence of antibacterial
R Derradi et al.
Langmuir : the ACS journal of surfaces and colloids, 35(47), 15232-15241 (2019-11-09)
Matrix vesicles (MVs) are a special class of extracellular vesicles that drive bone and dentin mineralization by providing the essential enzymes and ions for the nucleation and propagation of mineral crystals. Tissue-nonspecific alkaline phosphatase (TNAP) is an integral protein of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service