Skip to Content
Merck
All Photos(2)

Key Documents

810130C

Avanti

16:0-06:0 NBD PC

Avanti Research - A Croda Brand 810130C

Synonym(s):

1-palmitoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H62N5O11P
CAS Number:
Molecular Weight:
771.88
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810130C-1mg)
pkg of 5 × 1 mL (810130C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810130C

concentration

1 mg/mL (810130C-1mg)
1 mg/mL (810130C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphocholine is considered as a precursor molecule. It is formed during the breakdown of phosphatidylcholine (PC) metabolism. 16:0-06:0 NBD PC is a fluorescent lipid.

Application

16:0-06:0 NBD PC I suitable for use:
  • in the preparation of fluid phase fluorescent bilayers
  • to prepare liposomes/small unilamellar vesicles (SUVs)
  • as substrate in in vitro enzyme assay of lipoteichoic acid (LTA) synthase enzyme (LtaS) to determine the enzyme specificity

Packaging

5 mL Amber Glass Screw Cap Vial (810130C-1mg)
5 mL Amber Glass Screw Cap Vial (810130C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biology-cancer metabolic phenotype
Culf MC
NMR Metabolomics in Cancer Research, 59, 15-138 (2013)
Role of the biomolecular interactions in the structure and tribological properties of synovial fluid
Mirea DA, et al.
Tribology International, 59, 302-311 (2013)
Osmotic stress adaptation in Lactobacillus casei BL23 leads tostructural changes in the cell wall polymer lipoteichoic acid
Palomino MM, et al.
Microbiology, 159(11), 2416-2426 (2013)
Kevin N Baumann et al.
ACS nano, 14(2), 2316-2323 (2020-01-25)
The self-assembly of the protein clathrin on biological membranes facilitates essential processes of endocytosis and has provided a source of inspiration for materials design by the highly ordered structural appearance. By mimicking the architecture of the protein building blocks and
Alexander Ohmann et al.
Nature communications, 9(1), 2426-2426 (2018-06-23)
Mimicking enzyme function and increasing performance of naturally evolved proteins is one of the most challenging and intriguing aims of nanoscience. Here, we employ DNA nanotechnology to design a synthetic enzyme that substantially outperforms its biological archetypes. Consisting of only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service