Skip to Content
Merck
All Photos(2)

Documents

810133P

Avanti

18:1-12:0 NBD PC

Avanti Research - A Croda Brand 810133P, powder

Synonym(s):

1-Oleoyl-2-[12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl]-sn-Glycero-3-Phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H76N5O11P
CAS Number:
Molecular Weight:
882.07
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810133P-1mg)
pkg of 5 × 1 mg (810133P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810133P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphocholine is considered as a precursor molecule. It is formed during the breakdown of phosphatidylcholine metabolism.

Application

18:1-12:0 NBD PC is suitable for use:
  • as an optical probe of dynamics in the preparation of liposomes
  • as the fluorophore in the formation of neuropathy target esterase (NTE) catalytic domain (NEST) proteoliposome
  • as a component in bilayer membranes
  • to prepare lipid vesicles

Packaging

5 mL Amber Glass Screw Cap Vial (810133P-1mg)
5 mL Amber Glass Screw Cap Vial (810133P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antimicrobial peptides temporins B and L induce formation of tubular lipid protrusions from supported phospholipid bilayers
Domanov YA and Kinnunen PKJ
Biophysical Journal, 91(12), 4427-4439 (2006)
Comparison of liposomes formed by sonication and extrusion: rotational and translational diffusion of an embedded chromophore
Lapinski MM, et al.
Langmuir, 23(23), 11677-11683 (2007)
Biology-cancer metabolic phenotype
NMR Metabolomics in Cancer Research, 15-138 (2013)
Influence of lysophospholipid hydrolysis by the catalytic domain of neuropathy target esterase on the fluidity of bilayer lipid membranes
Greiner AJ, et al.
Biochimica et Biophysica Acta - Biomembranes, 1798(8), 1533-1539 (2010)
Haijia Yu et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(16), 4362-4367 (2016-04-05)
Organelles are in constant communication with each other through exchange of proteins (mediated by trafficking vesicles) and lipids [mediated by both trafficking vesicles and lipid transfer proteins (LTPs)]. It has long been known that vesicle trafficking can be tightly regulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service