Skip to Content
Merck
All Photos(2)

Key Documents

840475P

Avanti

18:1 (Δ9-Cis) PG

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); DOPG; PG(18:1(9Z)/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H78O10PNa
CAS Number:
Molecular Weight:
797.03
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dioleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840475P-1g)
pkg of 2 × 100 mg (840475P-200mg)
pkg of 5 × 100 mg (840475P-500mg)
pkg of 1 × 25 mg (840475P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C42H79O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,39-40,43-44H,3-16,21-38H2,1-2H3,(H,47,48);/q;+1/p-1/b19-17-,20-18-;/t39?,40-;/m

InChI key

IUVFCFQZFCOKRC-IPKKNMRRSA-M

General description

Phosphoglyceride (PG) are also referred as glycerophospholipid. It is an important component of membrane bilayers. It has three-carbon backbone of glycerol, two long-chain fatty acids, esterified to hydroxyl groups on carbons 1 and 2 of the glycerol and phosphoric acid esterified to the C3 hydroxyl group of glycerol. It is found at high levels in the membranes of all cells and at low levels in fat stores. 1,2-dioleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (DOPG)/18:1 (Δ9-Cis) PG has two hydroxyl groups in its head group. It is an anionic lipid.

Application

18:1 (Δ9-Cis) PG has been used:
  • in the preparation of giant unilamellar vesicle (GUV)
  • in the preparation 200 nm large unilamellar vesicles (LUV)
  • as a supplement in native macrophage membranes to generate membrane liposomes with an artificially modified electrical charge

Biochem/physiol Actions

Phosphoglycerides (PG)/glycerophospholipids play a key role in cell signaling systems. It also serves as an anchor for proteins in cell membranes.

Packaging

5 mL Clear Glass Sealed Ampule (840475P-200mg)
5 mL Clear Glass Sealed Ampule (840475P-25mg)
5 mL Clear Glass Sealed Ampule (840475P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (840475P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipids
Medical Biochemistry, 99- 119 (2017)
Membrane Structure and Dynamics
Cell Biology, 227- 239 (2017)
Isoeugenol has a non-disruptive detergent-like mechanism of action
Hyldgaard M, et al.
Frontiers in Microbiology, 6, 754-754 (2015)
Tripartite Chimeras Comprising Functional Domains Derived from the Cytosolic NADPH Oxidase Components p47phox, p67phox, and Rac1 Elicit Activator-independent Superoxide Production by Phagocyte Membranes AN ESSENTIAL ROLE FOR ANIONIC MEMBRANE PHOSPHOLIPIDS
Berdichevsky Y, et al.
The Journal of biological chemistry, 282(30), 22122- 22139 (2007)
Morten Hyldgaard et al.
Frontiers in microbiology, 6, 754-754 (2015-08-19)
Isoeugenol is an essential oil constituent of nutmeg, clove, and cinnamon. Despite isoeugenol's promising antimicrobial activity, no studies have yet investigated its mode of antibacterial action at the molecular level. The aim of this study is to clarify isoeugenol's antibacterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service