Skip to Content
Merck
All Photos(2)

Key Documents

850457C

Avanti

16:0-18:1 PC

Avanti Research - A Croda Brand

Synonym(s):

1-hexadecanoyl-2--(9Z-octadecenoyl)-sn-glycero-3-phosphocholine; POPC; PC(16:0/18:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO8P
CAS Number:
Molecular Weight:
760.08
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

description

1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850457C-25mg)
pkg of 2 × 20 mL (850457C-1g)
pkg of 2 × 4 mL (850457C-200mg)
pkg of 5 × 4 mL (850457C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (850457C-25mg)
25 mg/mL (850457C-1g)
25 mg/mL (850457C-200mg)
25 mg/mL (850457C-500mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20-/t40-/m1/s1

InChI key

WTJKGGKOPKCXLL-VYOBOKEXSA-N

General description

POPC is typically considered one of the model lipids for biophysical experiments. The fatty acid composition, i.e., saturated chain in the sn-1 position and unsaturated chain in the sn-2 position, mimics mammalian phospholipid composition. Since the major constituent in Egg PC is 1-palmitoyl-2-oleoyl PC, POPC is an excellent synthetic substitute for EPC.

Application

16:0-18:1 PC has been used in liposome disruption assay to examine lytic properties of biomolecules.[1][2] It has also been used to prepare liposomes.[3]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850457C-1g)
5 mL Clear Glass Sealed Ampule (850457C-200mg)
5 mL Clear Glass Sealed Ampule (850457C-25mg)
5 mL Clear Glass Sealed Ampule (850457C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liposome Disruption Assay to Examine Lytic Properties of Biomolecules.
Jimah JR, et al.
Bio-protocol, 7(15) (2017)
Membrane lipid screen to identify molecular targets of biomolecules
Jimah JR, et al.
Bio-protocol, 7(15) (2017)
Ionization Properties of Phospholipids Determined by Zeta Potential Measurements
Sathappa M, et al.
Bio-protocol, 6(22) (2016)
Tripta Bhatia et al.
ACS nano, 12(5), 4478-4485 (2018-04-17)
Giant unilamellar vesicles (GUVs) provide a direct connection between the nano- and the microregime. On the one hand, these vesicles represent biomimetic compartments with linear dimensions of many micrometers. On the other hand, the vesicle walls are provided by single
Michael J Hallock et al.
Biochemistry, 57(50), 6897-6905 (2018-11-21)
The plasma membrane of the cell is a complex, tightly regulated, heterogeneous environment shaped by proteins, lipids, and small molecules. Ca2+ ions are important cellular messengers, spatially separated from anionic lipids. After cell injury, disease, or apoptotic events, anionic lipids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service