Skip to Content
Merck
All Photos(2)

Key Documents

700059P

Avanti

27-hydroxycholesterol-d6

Avanti Research - A Croda Brand

Synonym(s):

25,26,26,26,27,27-hexadeuterocholest-5-ene-3β,27-diol; 27-hydroxycholest-5-en-3-ol(d6); 111011

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40D6O2
CAS Number:
Molecular Weight:
408.69
UNSPSC Code:
41141804
NACRES:
NA.25

description

25,26,26,26,27,27-hexadeuterocholest-5-ene-3β,27-diol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700059P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC[C@@]([2H])(C([2H])([2H])[2H])C(O)([2H])[2H]

General description

27-hydroxycholesterol (27-HC) is synthesized from cholesterol by the action of sterol 27-hydroxylase in the liver. It is an abundant oxysterol in the circulation ranging from 0.15 to 0.73 μM. 27-HC serves as a substrate for bile synthesis. 27-hydroxycholesterol-d6 is a deuterated form of 27-hydroxycholesterol.

Application

27-hydroxycholesterol-d6 has been used:
  • as an internal standard in isotope dilution-mass spectrometry analysis of heart mitochondria samples
  • as an internal standard in gas chromatography-mass spectrometry (GC–MS) for plasma sterol quantification
  • as a deuterated internal standard for spiking plasma samples for liquid chromatography with tandem mass spectrometry (LC-MS-MS) analysis

Biochem/physiol Actions

27-hydroxycholesterol (27-HC) is a weak liver X receptor (LXR) agonist and a selective estrogen receptor modulator (SERM). Elevated levels of 27-HC is observed in hypercholesterolemia. 27-HC is implicated in breast cancer tumor and glioblastoma progression. 27-HC may help in treating non-alcoholic steatohepatitis. Elevated levels of 27-HC is associated with atherosclerotic lesions.

Packaging

5 mL Amber Glass Screw Cap Vial (700059P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Liu et al.
Oncology letters, 18(4), 3623-3629 (2019-10-04)
Glioblastoma is the most frequent primary malignant brain tumor in adults. Oxysterols are oxidation products of cholesterol generated by enzymatic reactions. 27-hydroxycholesterol (27-HC), an oxysterol, is an abundant metabolite of cholesterol. 27-HC significantly accelerates mammary cancer growth, proliferation and progression
Irundika H K Dias et al.
Redox biology, 16, 139-145 (2018-03-04)
Oxysterols (OHC) are biologically active cholesterol metabolites circulating in plasma that may be formed enzymatically (e.g. 24S-OHC, 25-OHC and 27-OHC) or by autoxidative mechanisms (e.g. 7-ketocholesterol, 7β-OHC and 25-OHC). Oxysterols are more soluble than cholesterol and are reported to exert
Erik R Nelson et al.
Science (New York, N.Y.), 342(6162), 1094-1098 (2013-11-30)
Hypercholesterolemia is a risk factor for estrogen receptor (ER)-positive breast cancers and is associated with a decreased response of tumors to endocrine therapies. Here, we show that 27-hydroxycholesterol (27HC), a primary metabolite of cholesterol and an ER and liver X
Di Li et al.
Obesity (Silver Spring, Md.), 26(4), 713-722 (2018-02-25)
Although 27-hydroxycholesterol (27-HC) has been reported as a potent regulator of lipid homeostasis, its role in hepatic lipogenesis remains obscure. The present study was designed to investigate the impact of 27-HC on sterol regulatory element-binding protein 1 (SREBP-1) and hepatic
Shaneabbas Raza et al.
Cancer cell international, 17, 52-52 (2017-05-16)
For every six men, one will be diagnosed with prostate cancer (PCa) in their lifetime. Estrogen receptors (ERs) are known to play a role in prostate carcinogenesis. However, it is unclear whether the estrogenic effects are mediated by estrogen receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service