Skip to Content
Merck
All Photos(1)

Key Documents

LM4114

Avanti

27-hydroxycholesterol (D6)

Avanti Research - A Croda Brand

Synonym(s):

cholest-5-ene-3β,27-diol(d6)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H40D6O2
CAS Number:
Molecular Weight:
408.69
UNSPSC Code:
12352100
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM4114-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](O)C3)(C)C3=CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC([2H])(C(O)([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C27H46O2/c1-18(17-28)6-5-7-19(2)23-10-11-24-22-9-8-20-16-21(29)12-14-26(20,3)25(22)13-15-27(23,24)4/h8,18-19,21-25,28-29H,5-7,9-17H2,1-4H3/t18?,19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i1D3,17D2,18D

InChI key

FYHRJWMENCALJY-CDBHVEDQSA-N

General description

27-hydroxycholesterol is an abundant oxysterol in circulation.

Application

27-hydroxycholesterol (D6) or cholest-5-ene-3β,27-diol(d6) has been used as an internal standard to measure the concentration of 27-hydroxycholesterol in the pre- and post-statin-treated serum samples by mass spectrometry.

Biochem/physiol Actions

27-hydroxycholesterol is known to induce breast cancer cell proliferation and metastatic progression in experimental models. It might serve as a ligand for liver X receptors (LXR) and farnesoid X-activated receptors (FXRs). 27-hydroxycholesterol controls the action of hydrocymethylglutaryl-CoA reductase. It promotes cholesterol efflux from the vascular endothelium.

Packaging

2 mL Amber Glass Sealed Ampule (LM4114-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey Molendijk et al.
Biomolecules, 10(5) (2020-05-21)
Esophageal adenocarcinoma (EAC) incidence has been rapidly increasing, potentially associated with the prevalence of the risk factors gastroesophageal reflux disease (GERD), obesity, high-fat diet (HFD), and the precursor condition Barrett's esophagus (BE). EAC development occurs over several years, with stepwise
I Björkhem et al.
Journal of lipid research, 42(3), 366-371 (2001-03-20)
Infants with the cholesterol synthesis defect Smith- Lemli-Opitz syndrome (SLO) have reduced activity of the enzyme 7-dehydrocholesterol-7-reductase and accumulate 7-dehydrocholesterol, with the highest concentration in the brain. As a result of the generally reduced content of cholesterol, plasma levels of
Veera Venkata Ratnam Bandaru et al.
BMC neuroscience, 15, 137-137 (2014-12-30)
Cholesterol metabolism is important for the maintenance of myelin and neuronal membranes in the central nervous system. Blood concentrations of the brain specific cholesterol metabolite 24S-hydroxysterol to the peripheral metabolite 27-hydroxycholesterol may be useful surrogate markers for neurodegenerative diseases including
Nitai C Hait et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(3), 4293-4310 (2020-02-06)
Sphingosine kinase 2 (SphK2) is known to phosphorylate the nuclear sphingolipid metabolite to generate sphingosine-1-phosphate (S1P). Nuclear S1P is involved in epigenetic regulation of gene expression; however, the underlying mechanisms are not well understood. In this work, we have identified
Siker Kimbung et al.
Endocrine-related cancer, 24(7), 339-349 (2017-04-27)
The impact of systemic 27-hydroxycholesterol (27HC) and intratumoral CYP27A1 expression on pathobiology and clinical response to statins in breast cancer needs clarification. 27HC is an oxysterol produced from cholesterol by the monooxygenase CYP27A1, which regulates intracellular cholesterol homeostasis. 27HC also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service