Z675423
Replacement mesh inserts for mini-sieve micro sieve set
set of 1
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
Recommended Products
packaging
set of 1
manufacturer/tradename
Bel-Art F37846-0000
Related Categories
General description
Replacement mesh screens for Mini-Sieve micro sieve set. One each 25, 35, 45, 60, 80, 120, 170 and 230 standard mesh screens.
Choose from one of the most recent versions:
Certificates of Analysis (COA)