Skip to Content
Merck
All Photos(1)

Key Documents

W508454

Sigma-Aldrich

Acetovanillone

≥98%, FG

Synonym(s):

4′-Hydroxy-3′-methoxyacetophenone, Acetovanillone, Apocynin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)COCH3
CAS Number:
Molecular Weight:
166.17
Beilstein:
637373
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.142
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥98%

bp

263-265 °C/17 mmHg (lit.)

mp

112-115 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

vanilla

SMILES string

COc1cc(ccc1O)C(C)=O

InChI

1S/C9H10O3/c1-6(10)7-3-4-8(11)9(5-7)12-2/h3-5,11H,1-2H3

InChI key

DFYRUELUNQRZTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetovanillone is a structural analog of vanillin that occurs mainly in wine aged in oak barrels. It is a potent NADPH oxidase inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of volatile oak compounds in wine by headspace solid-phase microextraction and gas chromatography?mass spectrometry.
Carrillo JD, et al.
Journal of Chromatography A, 1102(1), 25-36 (2006)
Elham Khanicheh et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(9), 2187-2192 (2013-08-03)
Antioxidative drugs continue to be developed for the treatment of atherosclerosis. Apocynin is an nicotinamide adenine dinucleotide phosphate oxidase inhibitor with anti-inflammatory properties. We used contrast-enhanced ultrasound molecular imaging to assess whether short-term apocynin therapy in atherosclerosis reduces vascular oxidative
Jingfei Zhang et al.
Neuron, 82(1), 195-207 (2014-03-19)
Complement receptor 3 (CR3) activation in microglia is involved in neuroinflammation-related brain disorders and pruning of neuronal synapses. Hypoxia, often observed together with neuroinflammation in brain trauma, stroke, and neurodegenerative diseases, is thought to exacerbate inflammatory responses and synergistically enhance brain
Volatile composition of aged wine in used barrels of French oak and of American oak.
Cerdan TG, et al.
Food Research International, 35(7), 603-610 (2002)
Ji Chen et al.
Hypertension (Dallas, Tex. : 1979), 61(3), 681-689 (2012-12-26)
Angiotensin-converting enzyme 2 (ACE2) is a lately discovered enzyme catalyzing Angiotensin II into Angiotensin 1-7. Angiotensin II has been reported to impair endothelial progenitor cell (EPC) function and is detrimental to stroke. Here, we studied the role of ACE2 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service