Skip to Content
Merck
All Photos(1)

Key Documents

W381918

Sigma-Aldrich

L-Arginine

99%, FCC, FG

Synonym(s):

(S)-2-Amino-5-guanidinopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
174.20
FEMA Number:
3819
Beilstein:
1725413
EC Number:
MDL number:
UNSPSC Code:
12164502
eCl@ss:
32160406
PubChem Substance ID:
Flavis number:
17.003
NACRES:
NA.21
Organoleptic:
faint
grade:
FG
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 172.320

Assay

99%

form

powder or crystals

optical activity

[α]20/D +27°, c = 8 in 6 M HCl

mp

222 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

faint

SMILES string

N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1

InChI key

ODKSFYDXXFIFQN-BYPYZUCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

L-Arginine is an amino acid that plays a key role in many physiological processes such as tissue repair and reproduction. It is a key precursor for synthesizing nitric oxide in mammals. Due to these factors, the dietary supplementation with L-arginine may show a range of health benefits.

Application


  • Metabolomic approaches to dissect dysregulated metabolism in the progression of pre-diabetes to T2DM.: This study employs L-Arginine to examine metabolic changes during the transition from pre-diabetes to type 2 diabetes mellitus, offering new targets for early intervention (Ji et al., 2024 Jun). Link to the article.

  • Amino acid profile alteration in age-related atrial fibrillation.: Investigates the role of L-Arginine in cardiovascular aging, highlighting its potential in developing therapeutic strategies against atrial fibrillation associated with aging (Huang et al., 2024 Mar). Link to the article.

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L-Arginine Research Progress (2012)
Blood pressure and metabolic changes during dietary L-arginine supplementation in humans.
Siani A, et al.
American Journal of Hypertension, 13(5), 547-551 (2000)
Krishnan Suresh Kumar et al.
European journal of medicinal chemistry, 45(11), 5474-5479 (2010-08-21)
A new series of 3-(benzylideneamino)-2-phenylquinazoline-4(3H)-ones were prepared through Schiff base formation of 3-amino-2-phenyl quinazoline-4(3)H-one with various substituted carbonyl compounds. Their chemical structures were elucidated by spectral studies. Cytotoxicity and antiviral activity were evaluated against herpes simplex virus-1 (KOS), herpes simplex
Palaniraja Thandapani et al.
Molecular cell, 50(5), 613-623 (2013-06-12)
Motifs rich in arginines and glycines were recognized several decades ago to play functional roles and were termed glycine-arginine-rich (GAR) domains and/or RGG boxes. We review here the evolving functions of the RGG box along with several sequence variations that
Ahmed Moursy et al.
Nucleic acids research, 42(10), 6659-6672 (2014-04-03)
Regulation of SMN2 exon 7 splicing is crucial for the production of active SMN protein and the survival of Spinal Muscular Atrophy (SMA) patients. One of the most efficient activators of exon 7 inclusion is hnRNP G, which is recruited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service