Skip to Content
Merck
All Photos(1)

Documents

W380008

Sigma-Aldrich

4-Ethyloctanoic acid

≥98%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)(CH2)2CO2H
CAS Number:
Molecular Weight:
172.26
FEMA Number:
3800
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.079
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.439 (lit.)

bp

163 °C (lit.)

density

0.904 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty

SMILES string

CCCCC(CC)CCC(O)=O

InChI

1S/C10H20O2/c1-3-5-6-9(4-2)7-8-10(11)12/h9H,3-8H2,1-2H3,(H,11,12)

InChI key

PWKJMPFEQOHBAC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eri Iwata et al.
The Journal of veterinary medical science, 65(9), 1019-1021 (2003-10-09)
A major constituent of the characteristic "goaty odor" 4-ethyl octanoic acid (4EOA) was previously shown to have no primer pheromone activity. This was also confirmed by our own bioassay system utilizing the recording technique of neural activity of the hypothalamic
Yuping Liu et al.
Molecules (Basel, Switzerland), 15(8), 5104-5111 (2010-08-18)
Five sulfur-containing flavor compounds were synthesized for the first time by the reaction of 4-ethyloctanoyl chloride with sulfur-containing alcohols or mercaptans. The synthesized compounds are 3-(methylthio)propyl 4-ethyloctanoate, 2-methyl-3-tetrahydro-furanthiol 4-ethyloctanoate, 4-methyl-5-thiazoleethanol 4-ethyloctanoate, 2-furan-methanethiol 4-ethyloctanoate and 2-methyl-3-furanthiol 4-ethyloctanoate. These five synthetic sulfur-containing
Hardy Z Castada et al.
Rapid communications in mass spectrometry : RCM, 31(19), 1641-1650 (2017-07-29)
4-Methyloctanoic acid, 4-ethyloctanoic acid, 4-methylnonanoic acid, and 3-methylindole are primary contributors to the distinctive aroma and flavor of lamb meat. The reactions of H The chemical ionization of pure standards of 4-ethyloctanoic acid, 4-methyloctanoic acid, 4-ethylnonanoic acid, and 3-methylindole was
Philipp Denk et al.
Analytical and bioanalytical chemistry, 410(14), 3395-3404 (2018-04-07)
Elastic therapeutic tapes are an important tool in the field of physical therapy and medicine. These tapes contain types of adhesive. However, sensory evaluations revealed the release of pronounced and irritating odors of the tapes. Negative odors were, amongst others

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service