Skip to Content
Merck
All Photos(1)

Key Documents

W357502

Sigma-Aldrich

(±)-4-Methyloctanoic acid

≥98%, FG

Synonym(s):

4-methyl caprylic acid, Iso nonanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(CH3)CH2CH2CO2H
CAS Number:
Molecular Weight:
158.24
FEMA Number:
3575
EC Number:
Council of Europe no.:
11926
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.063
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥98%

refractive index

n20/D 1.433 (lit.)

bp

149 °C/22 mmHg (lit.)

density

0.91 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; dairy; fatty; milk; waxy

SMILES string

CCCCC(C)CCC(O)=O

InChI

1S/C9H18O2/c1-3-4-5-8(2)6-7-9(10)11/h8H,3-7H2,1-2H3,(H,10,11)

InChI key

LEGGANXCVQPIAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Methyloctanoic acid is a branched fatty acid commonly used in the flavor and fragrance industries. This compound gives sweaty and goaty flavors to the mutton meat and goat′s milk cheese, respectively.

Application

  • Medium-Chain Fatty Acids Rescue Motor Function and Neuromuscular Junction Degeneration in a Drosophila Model of Amyotrophic Lateral Sclerosis.: This research explores the therapeutic potential of medium-chain fatty acids, including 4-methyloctanoic acid, in rescuing motor function and preventing neuromuscular degeneration in a fruit fly model of ALS (Dunn et al., 2023).

  • Identification of muttony-related compounds in cooked mutton tallows and their flavor intensities subjected to phenolic extract from thyme (Thymus vulgaris L.).: The study identifies 4-methyloctanoic acid among other compounds responsible for the characteristic flavor of cooked mutton, demonstrating its impact on food flavor profiles (Wu et al., 2023).

  • Effects of Allium mongolicum Regel ethanol extract on three flavor-related rumen branched-chain fatty acids, rumen fermentation and rumen bacteria in lambs.: This paper investigates the effects of Allium mongolicum extract on rumen fermentation and bacteria in lambs, highlighting the role of 4-methyloctanoic acid in flavor formation (Zhao et al., 2022).

  • Identification and Inhibition of the Key Off-Odorants in Duck Broth by Means of the Sensomics Approach and Binary Odor Mixture.: This research identifies 4-methyloctanoic acid as a key off-odorant in duck broth and explores methods to inhibit its formation to improve food quality (Pu et al., 2022).

Biochem/physiol Actions

Odor at 1.0% PG
Taste at 10 ppm

Other Notes

Natural occurrence: Cooked mutton and lamb.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A novel method for determination and quantification of 4-methyloctanoic and 4-methylnonanoic acids in mutton by hollow fiber supported liquid membrane extraction coupled with gas chromatography
Chen H, et al.
Meat Science, 92(4), 715-720 (2012)
The effect of ethanol on the kinetics of lipase-mediated enantioselective esterification of 4-methyloctanoic acid and the hydrolysis of its ethyl ester
Heinsman NWJT, et al.
Biotechnology and Bioengineering, 76(3), 193-199 (2001)
Study of light-induced volatile compounds in goat's milk cheese
Kim G-Y, et al.
Journal of Agricultural and Food Chemistry, 51(5), 1405-1409 (2003)
Heidi Fleischer et al.
SLAS technology, 24(3), 330-341 (2019-01-09)
The monitoring of chemical reactions is an important task in chemical engineering, especially in quality control, pharmaceutical and biological processes, or industrial production. The development of new reactions such as catalyst-based procedures requires detailed knowledge about process steps and reaction
Hardy Z Castada et al.
Rapid communications in mass spectrometry : RCM, 31(19), 1641-1650 (2017-07-29)
4-Methyloctanoic acid, 4-ethyloctanoic acid, 4-methylnonanoic acid, and 3-methylindole are primary contributors to the distinctive aroma and flavor of lamb meat. The reactions of H The chemical ionization of pure standards of 4-ethyloctanoic acid, 4-methyloctanoic acid, 4-ethylnonanoic acid, and 3-methylindole was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service