Skip to Content
Merck
All Photos(1)

Key Documents

464430

Sigma-Aldrich

2-Butyloctanoic acid

96%

Synonym(s):

α-Butylcaprylic acid, 2-Butyl-caprylic acid, 2-Butyloctanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH[(CH2)3CH3]CO2H
CAS Number:
Molecular Weight:
200.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

96%

refractive index

n20/D 1.438 (lit.)

bp

230 °C (lit.)

density

0.887 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CCCCCCC(CCCC)C(O)=O

InChI

1S/C12H24O2/c1-3-5-7-8-10-11(12(13)14)9-6-4-2/h11H,3-10H2,1-2H3,(H,13,14)

InChI key

OARDBPIZDHVTCK-UHFFFAOYSA-N

Related Categories

General description

2-Butyloctanoic acid is a 2-butyl-substituted carboxylic acid.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aki Maruoka et al.
Biocontrol science, 25(2), 63-71 (2020-06-09)
House dust mites, Dermatophagoides pteronyssinus are present in the indoor environments, such as pillows and carpets. In this study, we investigated the mite control effect of branched chain fatty acids (2-ethylhexanoic acid (iso-C8), 2-butyloctanoic acid (iso-C12), isopalmitic acid (iso-C16) and
Overcrowding factors of mosquito larvae. VI. Structure-activity relations of 2-substituted aliphatic carboxylic acids against mosquito larvae.
Hwang Y-S, et al.
Journal of Agricultural and Food Chemistry, 22(6), 1004-1006 (1974)
Jumat Salimon et al.
Journal of automated methods & management in chemistry, 2011, 263624-263624 (2011-10-19)
A study was conveyed to produce estolide ester using ricinoleic acid as the backbone. The ricinoleic acid reacted with saturated fatty acid from C8-C18. These reactions were conducted under vacuum at 60°C for 24 h without solvent. The reaction used acid
Pishan Chang et al.
Neuropharmacology, 69, 105-114 (2012-11-28)
The medium chain triglyceride (MCT) ketogenic diet is used extensively for treating refractory childhood epilepsy. This diet increases the plasma levels of medium straight chain fatty acids. A role for these and related fatty acids in seizure control has not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service