Skip to Content
Merck
All Photos(1)

Key Documents

W284823

Sigma-Aldrich

Peppermint oil

purified by triple-distillation, FCC, FG

Synonym(s):

Mentha piperita

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2848
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

grade

FG
Kosher

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.230
FDA 21 CFR 182.20

optical activity

[α]20/D −26°, neat

purified by

triple-distillation

origin

USA origin

refractive index

n20/D 1.461 (lit.)

bp

215 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

peppermint

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M S Alam et al.
Mymensingh medical journal : MMJ, 22(1), 27-30 (2013-02-19)
Irritable bowel syndrome (IBS) is one of the most common functional gastrointestinal disorder which is associated with considerable sufferings of patient and Peppermint oil is volatile oil, its active principle is menthol-contain a cyclic monoterpine which has anti-spasmotic properties due
Elise Herro et al.
Dermatitis : contact, atopic, occupational, drug, 21(6), 327-329 (2010-12-15)
PEPPERMINT (Mentha piperita) is a popular herb that can be used in numerous forms (ie, oil, leaf, leaf extract, and leaf water). Peppermint oil has the most uses, and use data on the oil are considered relevant to the leaf
Paul Enck et al.
European journal of gastroenterology & hepatology, 22(12), 1402-1411 (2011-03-11)
Numerous meta-analyses have recently assessed the overall clinical benefit of single therapy options and groups of therapies in the irritable bowel syndrome (IBS). By large, this should enable physicians to select from a number of therapy options available. We entered
Hae Eun Park et al.
Journal of separation science, 35(3), 416-423 (2012-01-04)
The quantitative performance of a simple home-built preparative gas chromatography (prep-GC) arrangement was tested, incorporating a micro-fluidic Deans switch, with collection of the target compound in a deactivated uncoated capillary tube. Repeat injections of a standard solution and peppermint sample
A Shavakhi et al.
Acta gastro-enterologica Belgica, 75(3), 349-353 (2012-10-23)
Colonic spasm is an important problem in colonoscopy for endoscopists to advance the colonoscope and visualize the mucosa. In the present study, we evaluated the efficacy of enteric-coated peppermint oil capsules (Colpermin) as an orally administered antispasmodic premedication in colonoscopy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service