Skip to Content
Merck
All Photos(1)

Key Documents

W282510

Sigma-Aldrich

Orange oil

natural, cold-compressed, California origin, FG

Synonym(s):

Citrus aurantium, Citrus sinensis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2825
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
biological source:
Citrus sinensis (L.) Osbeck
food allergen:
no known allergens

biological source

Citrus sinensis (L.) Osbeck

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 872/2012

optical activity

[α]20/D +97°, neat

quality

cold-compressed

may contain

<0.02% BHT, synthetic as stabilizer

origin

California origin

refractive index

n20/D 1.473

bp

177 °C

density

0.843 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

orange

Looking for similar products? Visit Product Comparison Guide

General description

Orange oil has been used to study its influence as a semiochemical on the mating success of mature males of the natal fruit fly.

Preparation Note

Extraction method: cold pressed

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adult diet and exposure to semiochemicals influence male mating success in Ceratitis rosa (Diptera: Tephritidae).
Quilici S, et al.
Journal of Applied Entomology , 137(s1), 142-153 (2013)
Quantitative composition of cold-pressed orange oils.
Shaw PE & Coleman RL.
Journal of Agricultural and Food Chemistry, 22(5), 785-787 (1974)
Terpenes and Sesquiterpenes in Cold?pressed Orange Oil.
Hunter GLK & Brogden WB.
Journal of Food Science, 30(1), 1-4 (1965)
David Julian McClements et al.
Journal of food science, 77(1), C33-C38 (2011-12-03)
Beverage emulsions containing flavor oils that have a relatively high water-solubility are unstable to droplet growth due to Ostwald ripening. The aim of this study was to improve the stability of model beverage emulsions to this kind of droplet growth
Sean J Pendleton et al.
Journal of food science, 77(6), M308-M311 (2012-05-16)
Plant essential oils have previously been shown to exhibit antimicrobial activities against various microorganisms. In this study, cold pressed terpeneless Valencia orange oil (CPTVO) was examined at various temperatures (37, 10, and 4 °C) to determine its antimicrobial activity against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service